Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29572010-29572241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TP53I13
Entrez Gene ID
90313
Gene Name
tumor protein p53 inducible protein 13
Gene Aliases
DSCP1
UniGene
Hs.514050
Gene Chromosome Location
Chr.17: 29568067-29573157 on Build GRCh38
Transcripts
NM_138349.2 XM_017025282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3672397 c.547C>A p.R183R Chr.17: 29572175-29572175 on Build GRCh38
3515657 c.520G>A p.A174T Chr.17: 29572148-29572148 on Build GRCh38
5525704 c.473C>T p.T158I Chr.17: 29572017-29572017 on Build GRCh38
7123195 c.482G>C p.R161T Chr.17: 29572026-29572026 on Build GRCh38
4735473 c.550delC p.P185fs*5 Chr.17: 29572178-29572178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755022067 Chr.17: 29572200-29572200 on Build GRCh38
rs200357604 Chr.17: 29572175-29572175 on Build GRCh38
rs200220543 Chr.17: 29572166-29572166 on Build GRCh38
rs756444334 Chr.17: 29572114-29572114 on Build GRCh38
rs577200698 Chr.17: 29572176-29572176 on Build GRCh38
rs374274922 Chr.17: 29572167-29572167 on Build GRCh38
rs200636117 Chr.17: 29572116-29572116 on Build GRCh38
rs757079652 Chr.17: 29572044-29572044 on Build GRCh38
rs781570721 Chr.17: 29572046-29572046 on Build GRCh38
rs748199090 Chr.17: 29572209-29572209 on Build GRCh38
rs377286429 Chr.17: 29572073-29572073 on Build GRCh38
rs766538307 Chr.17: 29572193-29572193 on Build GRCh38
rs201515753 Chr.17: 29572092-29572092 on Build GRCh38
rs201997027 Chr.17: 29572082-29572082 on Build GRCh38
rs779289120 Chr.17: 29572125-29572125 on Build GRCh38
rs764842163 Chr.17: 29572089-29572089 on Build GRCh38
rs1008902741 Chr.17: 29572148-29572148 on Build GRCh38
rs200995959 Chr.17: 29572066-29572066 on Build GRCh38
rs1036788784 Chr.17: 29572197-29572197 on Build GRCh38
rs748716214 Chr.17: 29572134-29572134 on Build GRCh38
rs553644354 Chr.17: 29572112-29572112 on Build GRCh38
rs766162438 Chr.17: 29572028-29572028 on Build GRCh38
rs772567922 Chr.17: 29572136-29572136 on Build GRCh38
rs780298645 Chr.17: 29572115-29572115 on Build GRCh38
rs890491603 Chr.17: 29572142-29572142 on Build GRCh38
rs966837782 Chr.17: 29572187-29572187 on Build GRCh38
rs371528446 Chr.17: 29572155-29572155 on Build GRCh38
rs919246284 Chr.17: 29572218-29572218 on Build GRCh38
rs759857806 Chr.17: 29572190-29572190 on Build GRCh38
rs762112079 Chr.17: 29572087-29572087 on Build GRCh38
rs751500978 Chr.17: 29572060-29572061 on Build GRCh38
rs761908090 Chr.17: 29572177-29572177 on Build GRCh38
rs758310672 Chr.17: 29572030-29572030 on Build GRCh38
rs777746924 Chr.17: 29572040-29572040 on Build GRCh38
rs1047835048 Chr.17: 29572133-29572133 on Build GRCh38
rs752201101 Chr.17: 29572090-29572090 on Build GRCh38
rs928832569 Chr.17: 29572143-29572143 on Build GRCh38
rs200111294 Chr.17: 29572074-29572074 on Build GRCh38
rs754021169 Chr.17: 29572198-29572198 on Build GRCh38
rs762925909 Chr.17: 29572161-29572161 on Build GRCh38
rs779119855 Chr.17: 29572207-29572207 on Build GRCh38
rs754929004 Chr.17: 29572144-29572154 on Build GRCh38
rs556938990 Chr.17: 29572182-29572182 on Build GRCh38
rs372939319 Chr.17: 29572067-29572067 on Build GRCh38
rs544593578 Chr.17: 29572181-29572181 on Build GRCh38
rs535304874 Chr.17: 29572091-29572091 on Build GRCh38
rs781191186 Chr.17: 29572111-29572111 on Build GRCh38
rs534007903 Chr.17: 29572145-29572145 on Build GRCh38
rs751335473 Chr.17: 29572041-29572041 on Build GRCh38
rs773556961 Chr.17: 29572088-29572088 on Build GRCh38
rs746030203 Chr.17: 29572058-29572058 on Build GRCh38

View Full Product Details