Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 29101526-29101757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBA2
Entrez Gene ID
321
Gene Name
amyloid beta precursor protein binding family A member 2
Gene Aliases
D15S1518E, HsT16821, LIN-10, MGC:14091, MINT2, X11-BETA, X11L
UniGene
Hs.618112
Gene Chromosome Location
Chr.15: 28885479-29118315 on Build GRCh38
Transcripts
NM_001130414.1 NM_005503.3 XM_011521488.2 XM_011521489.2 XM_011521490.2 XM_011521491.1 XM_011521492.2 XM_011521493.2 XM_011521494.2 XM_011521495.1 XM_011521496.1 XM_017022101.1 XM_017022102.1 XM_017022103.1 XM_017022104.1 XM_017022105.1 XM_017022106.1 XM_017022107.1 XM_017022108.1 XM_017022109.1 XM_017022110.1 XM_017022111.1 XM_017022112.1 XM_017022113.1 XM_017022114.1 XM_017022115.1 XM_017022116.1 XM_017022117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3672014 c.1347G>T p.M449I Chr.15: 29101607-29101607 on Build GRCh38
2083870 c.1461G>A p.T487T Chr.15: 29101721-29101721 on Build GRCh38
7213268 c.1452C>T p.I484I Chr.15: 29101712-29101712 on Build GRCh38
147868 c.1460C>T p.T487M Chr.15: 29101720-29101720 on Build GRCh38
4972718 c.1405C>G p.L469V Chr.15: 29101665-29101665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142481200 Chr.15: 29101624-29101624 on Build GRCh38
rs753422882 Chr.15: 29101623-29101623 on Build GRCh38
rs773529837 Chr.15: 29101692-29101692 on Build GRCh38
rs750160692 Chr.15: 29101728-29101728 on Build GRCh38
rs573898603 Chr.15: 29101690-29101690 on Build GRCh38
rs978899574 Chr.15: 29101725-29101725 on Build GRCh38
rs899089675 Chr.15: 29101616-29101616 on Build GRCh38
rs371434692 Chr.15: 29101594-29101594 on Build GRCh38
rs763193966 Chr.15: 29101710-29101710 on Build GRCh38
rs774705861 Chr.15: 29101712-29101712 on Build GRCh38
rs763650417 Chr.15: 29101606-29101606 on Build GRCh38
rs757032213 Chr.15: 29101548-29101548 on Build GRCh38
rs768927880 Chr.15: 29101570-29101570 on Build GRCh38
rs778632122 Chr.15: 29101640-29101640 on Build GRCh38
rs202208854 Chr.15: 29101677-29101677 on Build GRCh38
rs776983304 Chr.15: 29101574-29101574 on Build GRCh38
rs745630621 Chr.15: 29101657-29101657 on Build GRCh38
rs750215090 Chr.15: 29101666-29101666 on Build GRCh38
rs572622006 Chr.15: 29101565-29101565 on Build GRCh38
rs761364766 Chr.15: 29101721-29101721 on Build GRCh38
rs779264391 Chr.15: 29101681-29101681 on Build GRCh38
rs139362409 Chr.15: 29101686-29101686 on Build GRCh38
rs763554780 Chr.15: 29101713-29101713 on Build GRCh38
rs1000230590 Chr.15: 29101691-29101691 on Build GRCh38
rs765627101 Chr.15: 29101584-29101584 on Build GRCh38
rs748337090 Chr.15: 29101689-29101689 on Build GRCh38
rs199757047 Chr.15: 29101724-29101724 on Build GRCh38
rs150891491 Chr.15: 29101683-29101683 on Build GRCh38
rs771392666 Chr.15: 29101711-29101711 on Build GRCh38
rs541634742 Chr.15: 29101575-29101575 on Build GRCh38
rs545399704 Chr.15: 29101554-29101554 on Build GRCh38
rs373242548 Chr.15: 29101593-29101593 on Build GRCh38
rs747030618 Chr.15: 29101567-29101567 on Build GRCh38
rs752146440 Chr.15: 29101596-29101596 on Build GRCh38
rs149908437 Chr.15: 29101558-29101558 on Build GRCh38
rs755574474 Chr.15: 29101599-29101599 on Build GRCh38
rs150027586 Chr.15: 29101720-29101720 on Build GRCh38

View Full Product Details