Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 88737381-88737650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM13A
Entrez Gene ID
10144
Gene Name
family with sequence similarity 13 member A
Gene Aliases
ARHGAP48, FAM13A1
UniGene
Hs.714536
Gene Chromosome Location
Chr.4: 88725954-89111398 on Build GRCh38
Transcripts
NM_001015045.2 NM_001265578.1 NM_001265579.1 NM_001265580.1 NM_014883.3 XM_005262681.2 XM_005262682.2 XM_005262683.2 XM_005262684.3 XM_006714057.3 XM_011531516.1 XM_011531517.1 XM_011531518.1 XM_011531519.2 XM_017007624.1 XM_017007625.1 XM_017007626.1 XM_017007627.1 XM_017007628.1 XM_017007629.1 XM_017007630.1 XM_017007631.1 XM_017007632.1 XM_017007633.1 XM_017007634.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6355708 c.2563-1G>C p.? Chr.4: 88737556-88737556 on Build GRCh38
3606955 c.1605G>A p.R535R Chr.4: 88737535-88737535 on Build GRCh38
4413952 c.1634A>T p.E545V Chr.4: 88737506-88737506 on Build GRCh38
2957660 c.1633G>C p.E545Q Chr.4: 88737507-88737507 on Build GRCh38
4126692 c.1623G>C p.Q541H Chr.4: 88737517-88737517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554318453 Chr.4: 88737617-88737617 on Build GRCh38
rs565503216 Chr.4: 88737469-88737469 on Build GRCh38
rs192863431 Chr.4: 88737405-88737405 on Build GRCh38
rs765225165 Chr.4: 88737537-88737537 on Build GRCh38
rs772599952 Chr.4: 88737601-88737601 on Build GRCh38
rs777362472 Chr.4: 88737558-88737558 on Build GRCh38
rs966159764 Chr.4: 88737618-88737618 on Build GRCh38
rs772366424 Chr.4: 88737476-88737476 on Build GRCh38
rs765225837 Chr.4: 88737547-88737547 on Build GRCh38
rs894758505 Chr.4: 88737408-88737408 on Build GRCh38
rs145874119 Chr.4: 88737555-88737555 on Build GRCh38
rs769078783 Chr.4: 88737458-88737458 on Build GRCh38
rs369437932 Chr.4: 88737589-88737589 on Build GRCh38
rs769190343 Chr.4: 88737596-88737596 on Build GRCh38
rs1036581130 Chr.4: 88737404-88737404 on Build GRCh38
rs1007194816 Chr.4: 88737460-88737460 on Build GRCh38
rs762329332 Chr.4: 88737466-88737466 on Build GRCh38
rs774847461 Chr.4: 88737464-88737464 on Build GRCh38
rs773522649 Chr.4: 88737605-88737605 on Build GRCh38
rs751234239 Chr.4: 88737577-88737577 on Build GRCh38
rs752244461 Chr.4: 88737603-88737604 on Build GRCh38
rs748459640 Chr.4: 88737600-88737600 on Build GRCh38
rs753916142 Chr.4: 88737507-88737507 on Build GRCh38
rs773575695 Chr.4: 88737486-88737486 on Build GRCh38
rs759560580 Chr.4: 88737477-88737477 on Build GRCh38
rs780373580 Chr.4: 88737437-88737437 on Build GRCh38
rs760929283 Chr.4: 88737606-88737606 on Build GRCh38
rs995149894 Chr.4: 88737428-88737428 on Build GRCh38
rs1043645766 Chr.4: 88737501-88737501 on Build GRCh38
rs760634703 Chr.4: 88737488-88737488 on Build GRCh38
rs368755056 Chr.4: 88737494-88737494 on Build GRCh38
rs1019544067 Chr.4: 88737620-88737620 on Build GRCh38
rs185417320 Chr.4: 88737450-88737450 on Build GRCh38
rs756905221 Chr.4: 88737578-88737578 on Build GRCh38
rs779579224 Chr.4: 88737599-88737599 on Build GRCh38
rs1009758149 Chr.4: 88737622-88737622 on Build GRCh38
rs766504123 Chr.4: 88737502-88737502 on Build GRCh38

View Full Product Details