Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43996027-43996264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC24
Entrez Gene ID
149473
Gene Name
coiled-coil domain containing 24
Gene Aliases
-
UniGene
Hs.632394
Gene Chromosome Location
Chr.1: 43991247-43996528 on Build GRCh38
Transcripts
NM_152499.2 XM_017000423.1 XM_017000424.1 XM_017000425.1 XM_017000426.1 XM_017000427.1 XM_017000428.1 XM_017000429.1 XM_017000430.1 XM_017000431.1 XM_017000432.1 XM_017000433.1 XM_017000434.1 XM_017000435.1 XM_017000436.1 XM_017000437.1 XM_017000438.1 XM_017000439.1 XM_017000440.1 XM_017000441.1 XM_017000442.1 XM_017000445.1 XM_017000446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4559854 c.820G>A p.G274S Chr.1: 43996056-43996056 on Build GRCh38
3934751 c.828G>A p.S276S Chr.1: 43996064-43996064 on Build GRCh38
3490058 c.817C>T p.R273* Chr.1: 43996053-43996053 on Build GRCh38
4669885 c.812G>A p.R271Q Chr.1: 43996048-43996048 on Build GRCh38
6622017 c.829G>A p.A277T Chr.1: 43996065-43996065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2301983 Chr.1: 43996150-43996150 on Build GRCh38
rs780280304 Chr.1: 43996097-43996097 on Build GRCh38
rs775021346 Chr.1: 43996155-43996155 on Build GRCh38
rs762483863 Chr.1: 43996064-43996064 on Build GRCh38
rs374227974 Chr.1: 43996084-43996084 on Build GRCh38
rs776646200 Chr.1: 43996055-43996055 on Build GRCh38
rs751821010 Chr.1: 43996179-43996179 on Build GRCh38
rs376940328 Chr.1: 43996047-43996047 on Build GRCh38
rs770664724 Chr.1: 43996143-43996143 on Build GRCh38
rs767150964 Chr.1: 43996077-43996077 on Build GRCh38
rs906216242 Chr.1: 43996098-43996098 on Build GRCh38
rs148185519 Chr.1: 43996152-43996152 on Build GRCh38
rs548237253 Chr.1: 43996219-43996219 on Build GRCh38
rs767026408 Chr.1: 43996079-43996079 on Build GRCh38
rs778744655 Chr.1: 43996212-43996212 on Build GRCh38
rs565152417 Chr.1: 43996093-43996093 on Build GRCh38
rs962399223 Chr.1: 43996088-43996088 on Build GRCh38
rs112173356 Chr.1: 43996106-43996106 on Build GRCh38
rs748112233 Chr.1: 43996058-43996058 on Build GRCh38
rs528510558 Chr.1: 43996213-43996213 on Build GRCh38
rs141661966 Chr.1: 43996054-43996054 on Build GRCh38
rs749188788 Chr.1: 43996208-43996208 on Build GRCh38
rs186460548 Chr.1: 43996183-43996183 on Build GRCh38
rs370451361 Chr.1: 43996139-43996139 on Build GRCh38
rs369147549 Chr.1: 43996051-43996051 on Build GRCh38
rs146193484 Chr.1: 43996074-43996074 on Build GRCh38
rs765722045 Chr.1: 43996163-43996163 on Build GRCh38
rs769813109 Chr.1: 43996115-43996115 on Build GRCh38
rs533932341 Chr.1: 43996200-43996200 on Build GRCh38
rs1029595474 Chr.1: 43996192-43996192 on Build GRCh38
rs758686308 Chr.1: 43996048-43996048 on Build GRCh38
rs750920322 Chr.1: 43996173-43996173 on Build GRCh38
rs748096909 Chr.1: 43996113-43996113 on Build GRCh38
rs752875006 Chr.1: 43996199-43996199 on Build GRCh38
rs143924931 Chr.1: 43996083-43996083 on Build GRCh38
rs530272003 Chr.1: 43996094-43996094 on Build GRCh38
rs766105051 Chr.1: 43996070-43996070 on Build GRCh38
rs1011841743 Chr.1: 43996075-43996075 on Build GRCh38
rs34677908 Chr.1: 43996174-43996174 on Build GRCh38
rs748110259 Chr.1: 43996189-43996206 on Build GRCh38
rs139890675 Chr.1: 43996063-43996063 on Build GRCh38
rs563979511 Chr.1: 43996116-43996116 on Build GRCh38
rs151061560 Chr.1: 43996149-43996149 on Build GRCh38
rs770744938 Chr.1: 43996211-43996211 on Build GRCh38
rs751833250 Chr.1: 43996101-43996101 on Build GRCh38
rs781482910 Chr.1: 43996108-43996108 on Build GRCh38
rs954058980 Chr.1: 43996196-43996196 on Build GRCh38
rs985971654 Chr.1: 43996218-43996218 on Build GRCh38
rs755322885 Chr.1: 43996180-43996180 on Build GRCh38
rs762285212 Chr.1: 43996161-43996161 on Build GRCh38

View Full Product Details