Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1508028-1508293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5K
Entrez Gene ID
51763
Gene Name
inositol polyphosphate-5-phosphatase K
Gene Aliases
PPS, SKIP
UniGene
Hs.632238
Gene Chromosome Location
Chr.17: 1494575-1516888 on Build GRCh38
Transcripts
NM_001135642.1 NM_016532.3 NM_130766.2 XM_005256683.2 XM_005256685.1 XM_005256686.1 XM_011523934.1 XM_011523936.2 XM_017024755.1 XM_017024756.1 XM_017024757.1 XM_017024758.1 XM_017024759.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6686184 c.406C>T p.R136W Chr.17: 1508147-1508147 on Build GRCh38
5385733 c.579G>A p.M193I Chr.17: 1508202-1508202 on Build GRCh38
7110439 c.616C>A p.R206R Chr.17: 1508165-1508165 on Build GRCh38
3672355 c.389G>T p.R130L Chr.17: 1508164-1508164 on Build GRCh38
6686188 c.403A>C p.N135H Chr.17: 1508150-1508150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755745268 Chr.17: 1508238-1508238 on Build GRCh38
rs267604751 Chr.17: 1508133-1508133 on Build GRCh38
rs764256868 Chr.17: 1508086-1508086 on Build GRCh38
rs765784226 Chr.17: 1508122-1508122 on Build GRCh38
rs895532912 Chr.17: 1508165-1508165 on Build GRCh38
rs750776057 Chr.17: 1508127-1508127 on Build GRCh38
rs751904711 Chr.17: 1508225-1508225 on Build GRCh38
rs760614205 Chr.17: 1508083-1508083 on Build GRCh38
rs141189913 Chr.17: 1508195-1508195 on Build GRCh38
rs145248842 Chr.17: 1508181-1508181 on Build GRCh38
rs1016370822 Chr.17: 1508052-1508052 on Build GRCh38
rs1029616234 Chr.17: 1508094-1508094 on Build GRCh38
rs367689627 Chr.17: 1508109-1508109 on Build GRCh38
rs1016792309 Chr.17: 1508204-1508204 on Build GRCh38
rs771733563 Chr.17: 1508265-1508265 on Build GRCh38
rs372212393 Chr.17: 1508164-1508164 on Build GRCh38
rs748831902 Chr.17: 1508145-1508145 on Build GRCh38
rs770937374 Chr.17: 1508146-1508146 on Build GRCh38
rs747889190 Chr.17: 1508139-1508139 on Build GRCh38
rs527269584 Chr.17: 1508194-1508194 on Build GRCh38
rs755551914 Chr.17: 1508141-1508141 on Build GRCh38
rs751067651 Chr.17: 1508091-1508091 on Build GRCh38
rs867572537 Chr.17: 1508261-1508261 on Build GRCh38
rs745853561 Chr.17: 1508156-1508156 on Build GRCh38
rs754260551 Chr.17: 1508095-1508095 on Build GRCh38
rs536504602 Chr.17: 1508060-1508060 on Build GRCh38
rs61733755 Chr.17: 1508138-1508138 on Build GRCh38
rs771881400 Chr.17: 1508173-1508173 on Build GRCh38
rs753334520 Chr.17: 1508245-1508245 on Build GRCh38
rs977699234 Chr.17: 1508166-1508166 on Build GRCh38
rs745850389 Chr.17: 1508260-1508260 on Build GRCh38
rs777300530 Chr.17: 1508142-1508142 on Build GRCh38
rs766810838 Chr.17: 1508222-1508222 on Build GRCh38
rs978981933 Chr.17: 1508114-1508114 on Build GRCh38
rs74437970 Chr.17: 1508258-1508258 on Build GRCh38
rs754669925 Chr.17: 1508201-1508202 on Build GRCh38
rs775784880 Chr.17: 1508072-1508072 on Build GRCh38
rs372228512 Chr.17: 1508071-1508071 on Build GRCh38
rs987391994 Chr.17: 1508063-1508063 on Build GRCh38
rs768863430 Chr.17: 1508273-1508273 on Build GRCh38
rs756896203 Chr.17: 1508247-1508247 on Build GRCh38
rs149998861 Chr.17: 1508168-1508168 on Build GRCh38
rs760997395 Chr.17: 1508178-1508178 on Build GRCh38
rs763286078 Chr.17: 1508217-1508217 on Build GRCh38
rs776721764 Chr.17: 1508189-1508189 on Build GRCh38
rs776840958 Chr.17: 1508274-1508274 on Build GRCh38
rs910842982 Chr.17: 1508084-1508084 on Build GRCh38
rs760494361 Chr.17: 1508271-1508271 on Build GRCh38
rs777307480 Chr.17: 1508240-1508240 on Build GRCh38

View Full Product Details