Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2153154-2153379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB26
Entrez Gene ID
25837
Gene Name
RAB26, member RAS oncogene family
Gene Aliases
V46133
UniGene
Hs.3797
Gene Chromosome Location
Chr.16: 2148144-2154165 on Build GRCh38
Transcripts
NM_001308053.1 NM_014353.4 XM_011522448.1 XM_011522450.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7386619 c.485G>A p.R162H Chr.16: 2153333-2153333 on Build GRCh38
6235092 c.409T>C p.F137L Chr.16: 2153161-2153161 on Build GRCh38
6538421 c.682C>G p.R228G Chr.16: 2153332-2153332 on Build GRCh38
5828358 c.668+1_668+2insT p.? Chr.16: 2153223-2153224 on Build GRCh38
6761326 c.723T>C p.H241H Chr.16: 2153373-2153373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756865696 Chr.16: 2153225-2153225 on Build GRCh38
rs560672272 Chr.16: 2153357-2153357 on Build GRCh38
rs977948819 Chr.16: 2153293-2153293 on Build GRCh38
rs189136439 Chr.16: 2153248-2153248 on Build GRCh38
rs758805899 Chr.16: 2153305-2153305 on Build GRCh38
rs996018558 Chr.16: 2153276-2153276 on Build GRCh38
rs147124016 Chr.16: 2153353-2153353 on Build GRCh38
rs779221883 Chr.16: 2153233-2153233 on Build GRCh38
rs575210074 Chr.16: 2153273-2153273 on Build GRCh38
rs751250523 Chr.16: 2153220-2153220 on Build GRCh38
rs771052489 Chr.16: 2153261-2153261 on Build GRCh38
rs200478430 Chr.16: 2153237-2153237 on Build GRCh38
rs763949549 Chr.16: 2153282-2153282 on Build GRCh38
rs192091078 Chr.16: 2153175-2153175 on Build GRCh38
rs772286953 Chr.16: 2153176-2153176 on Build GRCh38
rs756855734 Chr.16: 2153355-2153356 on Build GRCh38
rs760300677 Chr.16: 2153347-2153347 on Build GRCh38
rs377523491 Chr.16: 2153300-2153300 on Build GRCh38
rs751303619 Chr.16: 2153283-2153283 on Build GRCh38
rs761579332 Chr.16: 2153290-2153290 on Build GRCh38
rs745313974 Chr.16: 2153230-2153230 on Build GRCh38
rs974345936 Chr.16: 2153335-2153335 on Build GRCh38
rs183498688 Chr.16: 2153234-2153234 on Build GRCh38
rs199569631 Chr.16: 2153301-2153301 on Build GRCh38
rs759499711 Chr.16: 2153185-2153185 on Build GRCh38
rs778684237 Chr.16: 2153358-2153359 on Build GRCh38
rs778090802 Chr.16: 2153312-2153312 on Build GRCh38
rs749954442 Chr.16: 2153297-2153297 on Build GRCh38
rs765777706 Chr.16: 2153352-2153352 on Build GRCh38
rs778217092 Chr.16: 2153356-2153356 on Build GRCh38
rs763894444 Chr.16: 2153213-2153213 on Build GRCh38
rs149402189 Chr.16: 2153333-2153333 on Build GRCh38
rs773366929 Chr.16: 2153181-2153181 on Build GRCh38
rs1042423374 Chr.16: 2153334-2153334 on Build GRCh38
rs774317906 Chr.16: 2153339-2153339 on Build GRCh38
rs747203395 Chr.16: 2153250-2153250 on Build GRCh38
rs767237546 Chr.16: 2153296-2153296 on Build GRCh38
rs760803089 Chr.16: 2153183-2153183 on Build GRCh38
rs778013281 Chr.16: 2153275-2153276 on Build GRCh38
rs750032981 Chr.16: 2153344-2153344 on Build GRCh38
rs138858515 Chr.16: 2153184-2153184 on Build GRCh38
rs755521389 Chr.16: 2153299-2153299 on Build GRCh38
rs368805303 Chr.16: 2153229-2153229 on Build GRCh38
rs370574027 Chr.16: 2153314-2153314 on Build GRCh38
rs529533871 Chr.16: 2153358-2153358 on Build GRCh38
rs746015169 Chr.16: 2153319-2153319 on Build GRCh38
rs756453356 Chr.16: 2153223-2153224 on Build GRCh38
rs758069332 Chr.16: 2153202-2153202 on Build GRCh38
rs758930562 Chr.16: 2153355-2153355 on Build GRCh38
rs917018336 Chr.16: 2153188-2153188 on Build GRCh38
rs755541208 Chr.16: 2153231-2153231 on Build GRCh38
rs767213293 Chr.16: 2153343-2153343 on Build GRCh38
rs762960636 Chr.16: 2153328-2153328 on Build GRCh38
rs1017169831 Chr.16: 2153172-2153172 on Build GRCh38
rs752537050 Chr.16: 2153197-2153197 on Build GRCh38
rs775274406 Chr.16: 2153278-2153278 on Build GRCh38
rs577853654 Chr.16: 2153323-2153323 on Build GRCh38
rs776797391 Chr.16: 2153263-2153263 on Build GRCh38
rs780018086 Chr.16: 2153359-2153359 on Build GRCh38
rs753141566 Chr.16: 2153303-2153303 on Build GRCh38
rs112925114 Chr.16: 2153315-2153315 on Build GRCh38
rs778039492 Chr.16: 2153241-2153241 on Build GRCh38
rs753236340 Chr.16: 2153212-2153212 on Build GRCh38
rs748903909 Chr.16: 2153318-2153318 on Build GRCh38
rs930172417 Chr.16: 2153345-2153345 on Build GRCh38
rs775535783 Chr.16: 2153325-2153325 on Build GRCh38
rs200678863 Chr.16: 2153360-2153360 on Build GRCh38
rs200884856 Chr.16: 2153193-2153193 on Build GRCh38
rs759463725 Chr.16: 2153269-2153269 on Build GRCh38

View Full Product Details