Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5777696-5777961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR52N5
Entrez Gene ID
390075
Gene Name
olfactory receptor family 52 subfamily N member 5
Gene Aliases
OR11-62, OR52N5Q
UniGene
Hs.690207
Gene Chromosome Location
Chr.11: 5777634-5778667 on Build GRCh38
Transcripts
NM_001001922.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5598590 c.777C>T p.I259I Chr.11: 5777858-5777858 on Build GRCh38
6734829 c.889C>A p.L297I Chr.11: 5777746-5777746 on Build GRCh38
688313 c.815G>A p.R272H Chr.11: 5777820-5777820 on Build GRCh38
246262 c.814C>T p.R272C Chr.11: 5777821-5777821 on Build GRCh38
5671578 c.760A>G p.I254V Chr.11: 5777875-5777875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748837736 Chr.11: 5777752-5777752 on Build GRCh38
rs75104866 Chr.11: 5777937-5777937 on Build GRCh38
rs780412659 Chr.11: 5777740-5777740 on Build GRCh38
rs758804759 Chr.11: 5777737-5777737 on Build GRCh38
rs867121206 Chr.11: 5777800-5777800 on Build GRCh38
rs771965335 Chr.11: 5777783-5777783 on Build GRCh38
rs144294327 Chr.11: 5777831-5777831 on Build GRCh38
rs764044554 Chr.11: 5777791-5777791 on Build GRCh38
rs750691301 Chr.11: 5777735-5777735 on Build GRCh38
rs755572387 Chr.11: 5777755-5777757 on Build GRCh38
rs747587083 Chr.11: 5777741-5777741 on Build GRCh38
rs770423321 Chr.11: 5777753-5777753 on Build GRCh38
rs751902450 Chr.11: 5777926-5777926 on Build GRCh38
rs868152200 Chr.11: 5777755-5777755 on Build GRCh38
rs150823468 Chr.11: 5777796-5777796 on Build GRCh38
rs146427121 Chr.11: 5777861-5777861 on Build GRCh38
rs777367889 Chr.11: 5777749-5777749 on Build GRCh38
rs781765535 Chr.11: 5777821-5777821 on Build GRCh38
rs753144504 Chr.11: 5777899-5777900 on Build GRCh38
rs760657642 Chr.11: 5777788-5777788 on Build GRCh38
rs570162187 Chr.11: 5777816-5777816 on Build GRCh38
rs746841673 Chr.11: 5777875-5777875 on Build GRCh38
rs755500369 Chr.11: 5777820-5777820 on Build GRCh38
rs761890209 Chr.11: 5777798-5777798 on Build GRCh38
rs867329035 Chr.11: 5777848-5777848 on Build GRCh38
rs377440089 Chr.11: 5777866-5777866 on Build GRCh38
rs763224026 Chr.11: 5777811-5777811 on Build GRCh38
rs149998822 Chr.11: 5777817-5777817 on Build GRCh38
rs755656222 Chr.11: 5777742-5777742 on Build GRCh38
rs117500523 Chr.11: 5777908-5777908 on Build GRCh38
rs200980931 Chr.11: 5777907-5777907 on Build GRCh38
rs147846695 Chr.11: 5777914-5777914 on Build GRCh38
rs143355934 Chr.11: 5777795-5777795 on Build GRCh38
rs745571889 Chr.11: 5777853-5777853 on Build GRCh38
rs200685016 Chr.11: 5777888-5777888 on Build GRCh38
rs775257594 Chr.11: 5777785-5777785 on Build GRCh38
rs753340779 Chr.11: 5777822-5777822 on Build GRCh38
rs766538363 Chr.11: 5777921-5777921 on Build GRCh38
rs750715469 Chr.11: 5777809-5777809 on Build GRCh38
rs778422227 Chr.11: 5777844-5777844 on Build GRCh38
rs758910287 Chr.11: 5777905-5777910 on Build GRCh38
rs776566982 Chr.11: 5777891-5777891 on Build GRCh38
rs765969707 Chr.11: 5777810-5777810 on Build GRCh38
rs745632919 Chr.11: 5777781-5777781 on Build GRCh38
rs761786865 Chr.11: 5777898-5777898 on Build GRCh38
rs935907256 Chr.11: 5777845-5777845 on Build GRCh38
rs774066405 Chr.11: 5777776-5777776 on Build GRCh38
rs139255166 Chr.11: 5777814-5777814 on Build GRCh38
rs765318342 Chr.11: 5777803-5777803 on Build GRCh38
rs372078375 Chr.11: 5777934-5777934 on Build GRCh38
rs753427119 Chr.11: 5777933-5777933 on Build GRCh38

View Full Product Details