Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 227747656-227747915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAP47
Entrez Gene ID
116841
Gene Name
synaptosome associated protein 47
Gene Aliases
C1orf142, ESFI5812, HEL-S-290, HEL170, SNAP-47, SVAP1
UniGene
Hs.325081
Gene Chromosome Location
Chr.1: 227728168-227781231 on Build GRCh38
Transcripts
NM_001323930.1 NM_001323931.1 NM_001323932.1 NM_001323933.1 NM_001323934.1 NM_001323935.1 NM_053052.3 NR_136654.1 NR_136655.1 NR_136656.1 NR_136657.1 XM_006711734.3 XM_017000231.1 XM_017000232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7222608 c.142A>G p.R48G Chr.1: 227747743-227747743 on Build GRCh38
7287000 c.175T>C p.Y59H Chr.1: 227747776-227747776 on Build GRCh38
7264423 c.228G>A p.S76S Chr.1: 227747829-227747829 on Build GRCh38
3484363 c.96G>A p.Q32Q Chr.1: 227747697-227747697 on Build GRCh38
7222607 c.142A>G p.R48G Chr.1: 227747743-227747743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370000908 Chr.1: 227747870-227747870 on Build GRCh38
rs200475270 Chr.1: 227747825-227747825 on Build GRCh38
rs772915561 Chr.1: 227747848-227747848 on Build GRCh38
rs34910471 Chr.1: 227747769-227747769 on Build GRCh38
rs752146412 Chr.1: 227747876-227747876 on Build GRCh38
rs764032403 Chr.1: 227747746-227747746 on Build GRCh38
rs201328130 Chr.1: 227747770-227747770 on Build GRCh38
rs780903502 Chr.1: 227747706-227747706 on Build GRCh38
rs746053419 Chr.1: 227747735-227747735 on Build GRCh38
rs566588891 Chr.1: 227747707-227747707 on Build GRCh38
rs762844496 Chr.1: 227747693-227747693 on Build GRCh38
rs2236359 Chr.1: 227747743-227747743 on Build GRCh38
rs376478626 Chr.1: 227747845-227747845 on Build GRCh38
rs946246198 Chr.1: 227747777-227747777 on Build GRCh38
rs544590913 Chr.1: 227747881-227747881 on Build GRCh38
rs112717702 Chr.1: 227747838-227747838 on Build GRCh38
rs369634626 Chr.1: 227747676-227747676 on Build GRCh38
rs575875893 Chr.1: 227747747-227747747 on Build GRCh38
rs749376866 Chr.1: 227747832-227747832 on Build GRCh38
rs761635991 Chr.1: 227747844-227747844 on Build GRCh38
rs910069842 Chr.1: 227747817-227747817 on Build GRCh38
rs763981209 Chr.1: 227747694-227747694 on Build GRCh38
rs756989237 Chr.1: 227747700-227747700 on Build GRCh38
rs140225131 Chr.1: 227747829-227747829 on Build GRCh38
rs776646959 Chr.1: 227747675-227747675 on Build GRCh38
rs778364082 Chr.1: 227747717-227747717 on Build GRCh38
rs756436802 Chr.1: 227747882-227747882 on Build GRCh38
rs768719759 Chr.1: 227747834-227747834 on Build GRCh38
rs780323476 Chr.1: 227747888-227747888 on Build GRCh38
rs747504970 Chr.1: 227747803-227747803 on Build GRCh38
rs769955830 Chr.1: 227747741-227747741 on Build GRCh38
rs757689741 Chr.1: 227747812-227747812 on Build GRCh38
rs912975166 Chr.1: 227747774-227747774 on Build GRCh38
rs747691698 Chr.1: 227747793-227747793 on Build GRCh38
rs527419212 Chr.1: 227747718-227747718 on Build GRCh38
rs150812885 Chr.1: 227747720-227747720 on Build GRCh38
rs761737665 Chr.1: 227747755-227747755 on Build GRCh38
rs757734576 Chr.1: 227747878-227747878 on Build GRCh38
rs746201803 Chr.1: 227747826-227747826 on Build GRCh38
rs557894284 Chr.1: 227747797-227747797 on Build GRCh38
rs202095982 Chr.1: 227747742-227747742 on Build GRCh38
rs373320394 Chr.1: 227747715-227747715 on Build GRCh38
rs751379759 Chr.1: 227747697-227747697 on Build GRCh38
rs760456187 Chr.1: 227747853-227747853 on Build GRCh38
rs774333210 Chr.1: 227747835-227747835 on Build GRCh38
rs58295986 Chr.1: 227747716-227747716 on Build GRCh38
rs941500599 Chr.1: 227747820-227747820 on Build GRCh38
rs752678571 Chr.1: 227747687-227747687 on Build GRCh38
rs767207634 Chr.1: 227747768-227747768 on Build GRCh38
rs766130401 Chr.1: 227747859-227747859 on Build GRCh38
rs776969353 Chr.1: 227747731-227747731 on Build GRCh38
rs765218060 Chr.1: 227747682-227747682 on Build GRCh38
rs386639987 Chr.1: 227747716-227747718 on Build GRCh38
rs145005377 Chr.1: 227747798-227747798 on Build GRCh38
rs201223656 Chr.1: 227747725-227747725 on Build GRCh38
rs961236254 Chr.1: 227747685-227747685 on Build GRCh38
rs151133242 Chr.1: 227747828-227747828 on Build GRCh38
rs753410063 Chr.1: 227747796-227747796 on Build GRCh38
rs201578220 Chr.1: 227747869-227747869 on Build GRCh38

View Full Product Details