Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 126631636-126631899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX43
Entrez Gene ID
389320
Gene Name
testis expressed 43
Gene Aliases
C5orf48, Tseg7
UniGene
Hs.177983
Gene Chromosome Location
Chr.5: 126631722-126636282 on Build GRCh38
Transcripts
NM_207408.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1198902 c.67C>A p.L23I Chr.5: 126631801-126631801 on Build GRCh38
3776137 c.69C>G p.L23L Chr.5: 126631803-126631803 on Build GRCh38
3776136 c.5C>A p.A2D Chr.5: 126631739-126631739 on Build GRCh38
1130951 c.35C>G p.P12R Chr.5: 126631769-126631769 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758055986 Chr.5: 126631725-126631725 on Build GRCh38
rs908631891 Chr.5: 126631860-126631860 on Build GRCh38
rs994657988 Chr.5: 126631749-126631749 on Build GRCh38
rs978904310 Chr.5: 126631690-126631690 on Build GRCh38
rs6862980 Chr.5: 126631712-126631712 on Build GRCh38
rs767906214 Chr.5: 126631686-126631686 on Build GRCh38
rs374235269 Chr.5: 126631813-126631813 on Build GRCh38
rs369820165 Chr.5: 126631713-126631713 on Build GRCh38
rs773412306 Chr.5: 126631800-126631801 on Build GRCh38
rs1020041201 Chr.5: 126631779-126631779 on Build GRCh38
rs144489109 Chr.5: 126631808-126631808 on Build GRCh38
rs773527059 Chr.5: 126631845-126631845 on Build GRCh38
rs761844346 Chr.5: 126631831-126631831 on Build GRCh38
rs750217421 Chr.5: 126631701-126631701 on Build GRCh38
rs139451988 Chr.5: 126631711-126631711 on Build GRCh38
rs779929639 Chr.5: 126631743-126631743 on Build GRCh38
rs941151440 Chr.5: 126631874-126631874 on Build GRCh38
rs185339208 Chr.5: 126631673-126631673 on Build GRCh38
rs746204229 Chr.5: 126631803-126631803 on Build GRCh38
rs766307457 Chr.5: 126631707-126631707 on Build GRCh38
rs760587098 Chr.5: 126631706-126631706 on Build GRCh38
rs889886614 Chr.5: 126631757-126631757 on Build GRCh38
rs1006853699 Chr.5: 126631769-126631769 on Build GRCh38
rs988336303 Chr.5: 126631665-126631665 on Build GRCh38
rs71587904 Chr.5: 126631839-126631839 on Build GRCh38
rs752217439 Chr.5: 126631718-126631718 on Build GRCh38
rs777451700 Chr.5: 126631751-126631751 on Build GRCh38
rs376995857 Chr.5: 126631870-126631870 on Build GRCh38
rs146677490 Chr.5: 126631814-126631814 on Build GRCh38
rs760820724 Chr.5: 126631860-126631861 on Build GRCh38
rs772388417 Chr.5: 126631832-126631832 on Build GRCh38
rs766472150 Chr.5: 126631865-126631882 on Build GRCh38
rs769107257 Chr.5: 126631821-126631821 on Build GRCh38
rs906414216 Chr.5: 126631666-126631666 on Build GRCh38
rs563209877 Chr.5: 126631871-126631871 on Build GRCh38
rs760998722 Chr.5: 126631855-126631855 on Build GRCh38
rs867227596 Chr.5: 126631759-126631759 on Build GRCh38
rs774472829 Chr.5: 126631828-126631828 on Build GRCh38
rs770108067 Chr.5: 126631805-126631805 on Build GRCh38

View Full Product Details