Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 117172524-117172757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO21
Entrez Gene ID
23014
Gene Name
F-box protein 21
Gene Aliases
FBX21
UniGene
Hs.740376
Gene Chromosome Location
Chr.12: 117143473-117190500 on Build GRCh38
Transcripts
NM_015002.2 NM_033624.2 XM_017019037.1 XM_017019038.1 XM_017019039.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
120891 c.910A>T p.M304L Chr.12: 117172574-117172574 on Build GRCh38
3811154 c.938G>T p.R313L Chr.12: 117172546-117172546 on Build GRCh38
691897 c.894A>T p.T298T Chr.12: 117172590-117172590 on Build GRCh38
6485126 c.946G>T p.G316* Chr.12: 117172538-117172538 on Build GRCh38
5722045 c.901C>T p.P301S Chr.12: 117172583-117172583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904324904 Chr.12: 117172661-117172661 on Build GRCh38
rs750940430 Chr.12: 117172549-117172549 on Build GRCh38
rs745333232 Chr.12: 117172646-117172646 on Build GRCh38
rs755171358 Chr.12: 117172580-117172580 on Build GRCh38
rs758605968 Chr.12: 117172555-117172555 on Build GRCh38
rs766559914 Chr.12: 117172574-117172574 on Build GRCh38
rs146125510 Chr.12: 117172595-117172595 on Build GRCh38
rs768909747 Chr.12: 117172690-117172690 on Build GRCh38
rs775177256 Chr.12: 117172652-117172652 on Build GRCh38
rs770266905 Chr.12: 117172659-117172659 on Build GRCh38
rs777737876 Chr.12: 117172610-117172610 on Build GRCh38
rs765884481 Chr.12: 117172546-117172546 on Build GRCh38
rs943248424 Chr.12: 117172667-117172667 on Build GRCh38
rs142755416 Chr.12: 117172625-117172625 on Build GRCh38
rs75054040 Chr.12: 117172653-117172653 on Build GRCh38
rs781430402 Chr.12: 117172587-117172587 on Build GRCh38
rs918946868 Chr.12: 117172547-117172547 on Build GRCh38
rs751593156 Chr.12: 117172575-117172575 on Build GRCh38
rs558374918 Chr.12: 117172670-117172670 on Build GRCh38
rs555590450 Chr.12: 117172542-117172542 on Build GRCh38
rs910297059 Chr.12: 117172598-117172598 on Build GRCh38
rs376887336 Chr.12: 117172590-117172590 on Build GRCh38
rs932954933 Chr.12: 117172572-117172572 on Build GRCh38
rs771652184 Chr.12: 117172651-117172651 on Build GRCh38
rs578016266 Chr.12: 117172721-117172721 on Build GRCh38
rs755871921 Chr.12: 117172597-117172597 on Build GRCh38

View Full Product Details