Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54311251-54311502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LILRA5
Entrez Gene ID
353514
Gene Name
leukocyte immunoglobulin like receptor A5
Gene Aliases
CD85, CD85F, ILT-11, ILT11, LILRB7, LIR-9, LIR9
UniGene
Hs.710986
Gene Chromosome Location
Chr.19: 54307070-54313139 on Build GRCh38
Transcripts
NM_021250.3 NM_181879.2 NM_181985.3 NM_181986.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3707386 c.672A>G p.V224V Chr.19: 54311454-54311454 on Build GRCh38
6206699 c.675G>A p.W225* Chr.19: 54311451-54311451 on Build GRCh38
3363011 c.649T>G p.S217A Chr.19: 54311477-54311477 on Build GRCh38
6217584 c.732C>T p.F244F Chr.19: 54311394-54311394 on Build GRCh38
1001111 c.653G>A p.R218H Chr.19: 54311473-54311473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770092636 Chr.19: 54311354-54311354 on Build GRCh38
rs780113187 Chr.19: 54311373-54311373 on Build GRCh38
rs770585865 Chr.19: 54311418-54311418 on Build GRCh38
rs758114748 Chr.19: 54311325-54311325 on Build GRCh38
rs111554293 Chr.19: 54311472-54311472 on Build GRCh38
rs751434857 Chr.19: 54311335-54311335 on Build GRCh38
rs764999592 Chr.19: 54311422-54311422 on Build GRCh38
rs570831996 Chr.19: 54311301-54311301 on Build GRCh38
rs923722861 Chr.19: 54311429-54311429 on Build GRCh38
rs750547834 Chr.19: 54311456-54311456 on Build GRCh38
rs757346290 Chr.19: 54311338-54311338 on Build GRCh38
rs867451032 Chr.19: 54311463-54311463 on Build GRCh38
rs781073941 Chr.19: 54311454-54311454 on Build GRCh38
rs749404393 Chr.19: 54311474-54311474 on Build GRCh38
rs934952871 Chr.19: 54311436-54311436 on Build GRCh38
rs763743033 Chr.19: 54311449-54311449 on Build GRCh38
rs762351617 Chr.19: 54311413-54311413 on Build GRCh38
rs200277601 Chr.19: 54311313-54311313 on Build GRCh38
rs914727666 Chr.19: 54311408-54311408 on Build GRCh38
rs148309852 Chr.19: 54311421-54311421 on Build GRCh38
rs752288887 Chr.19: 54311423-54311423 on Build GRCh38
rs757148853 Chr.19: 54311450-54311450 on Build GRCh38
rs773246253 Chr.19: 54311288-54311288 on Build GRCh38
rs976116063 Chr.19: 54311290-54311290 on Build GRCh38
rs375499197 Chr.19: 54311473-54311473 on Build GRCh38
rs768898476 Chr.19: 54311388-54311388 on Build GRCh38
rs781202835 Chr.19: 54311340-54311340 on Build GRCh38
rs931602565 Chr.19: 54311336-54311336 on Build GRCh38
rs777837192 Chr.19: 54311331-54311331 on Build GRCh38
rs746118070 Chr.19: 54311349-54311349 on Build GRCh38
rs753557228 Chr.19: 54311302-54311302 on Build GRCh38
rs775002598 Chr.19: 54311389-54311389 on Build GRCh38
rs764767573 Chr.19: 54311314-54311314 on Build GRCh38
rs375283642 Chr.19: 54311384-54311384 on Build GRCh38
rs772158044 Chr.19: 54311481-54311481 on Build GRCh38
rs747553929 Chr.19: 54311372-54311372 on Build GRCh38
rs912327905 Chr.19: 54311275-54311275 on Build GRCh38
rs752587842 Chr.19: 54311318-54311318 on Build GRCh38
rs1056151262 Chr.19: 54311467-54311467 on Build GRCh38
rs141466672 Chr.19: 54311435-54311435 on Build GRCh38
rs765919097 Chr.19: 54311296-54311296 on Build GRCh38

View Full Product Details