Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 36598206-36598475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS11
Entrez Gene ID
79154
Gene Name
dehydrogenase/reductase 11
Gene Aliases
ARPG836, SDR24C1, spDHRS11
UniGene
Hs.462859
Gene Chromosome Location
Chr.17: 36591791-36600804 on Build GRCh38
Transcripts
NM_024308.3 XM_005257658.2 XM_011525233.2

SNP Detail

SNP ID Chromosome Location
rs529569640 Chr.17: 36598254-36598254 on Build GRCh38
rs751894376 Chr.17: 36598238-36598238 on Build GRCh38
rs772821875 Chr.17: 36598288-36598288 on Build GRCh38
rs753109449 Chr.17: 36598229-36598229 on Build GRCh38
rs775545621 Chr.17: 36598255-36598255 on Build GRCh38
rs778614206 Chr.17: 36598239-36598239 on Build GRCh38
rs371420429 Chr.17: 36598312-36598312 on Build GRCh38
rs188310364 Chr.17: 36598307-36598307 on Build GRCh38
rs556523812 Chr.17: 36598435-36598435 on Build GRCh38
rs571006516 Chr.17: 36598384-36598384 on Build GRCh38
rs370883504 Chr.17: 36598273-36598273 on Build GRCh38
rs892190695 Chr.17: 36598446-36598446 on Build GRCh38
rs921233384 Chr.17: 36598356-36598356 on Build GRCh38
rs180825246 Chr.17: 36598326-36598326 on Build GRCh38
rs536688112 Chr.17: 36598427-36598427 on Build GRCh38
rs771831893 Chr.17: 36598285-36598285 on Build GRCh38
rs987010163 Chr.17: 36598414-36598414 on Build GRCh38
rs146654963 Chr.17: 36598231-36598231 on Build GRCh38
rs771717340 Chr.17: 36598260-36598260 on Build GRCh38
rs769653287 Chr.17: 36598243-36598243 on Build GRCh38
rs1052612570 Chr.17: 36598412-36598412 on Build GRCh38
rs745706948 Chr.17: 36598244-36598244 on Build GRCh38
rs774026461 Chr.17: 36598298-36598299 on Build GRCh38
rs551063769 Chr.17: 36598382-36598382 on Build GRCh38
rs754969899 Chr.17: 36598262-36598262 on Build GRCh38
rs778518352 Chr.17: 36598267-36598267 on Build GRCh38
rs912184109 Chr.17: 36598278-36598278 on Build GRCh38
rs770382142 Chr.17: 36598295-36598296 on Build GRCh38
rs781468524 Chr.17: 36598241-36598241 on Build GRCh38
rs764362949 Chr.17: 36598232-36598232 on Build GRCh38
rs912860151 Chr.17: 36598423-36598423 on Build GRCh38

View Full Product Details