Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 118749579-118749852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL13RA1
Entrez Gene ID
3597
Gene Name
interleukin 13 receptor subunit alpha 1
Gene Aliases
CD213A1, CT19, IL-13Ra, NR4
UniGene
Hs.496646
Gene Chromosome Location
Chr.X: 118726954-118794533 on Build GRCh38
Transcripts
NM_001560.2 XM_011531336.1 XM_017029507.1

SNP Detail

SNP ID Chromosome Location
rs899553053 Chr.X: 118749786-118749786 on Build GRCh38
rs940748374 Chr.X: 118749768-118749768 on Build GRCh38
rs761476755 Chr.X: 118749763-118749763 on Build GRCh38
rs754036451 Chr.X: 118749826-118749826 on Build GRCh38
rs747126033 Chr.X: 118749713-118749713 on Build GRCh38
rs1051164712 Chr.X: 118749797-118749797 on Build GRCh38
rs763292056 Chr.X: 118749791-118749791 on Build GRCh38
rs145506768 Chr.X: 118749717-118749717 on Build GRCh38
rs927360198 Chr.X: 118749602-118749602 on Build GRCh38
rs138018049 Chr.X: 118749761-118749761 on Build GRCh38
rs776200896 Chr.X: 118749823-118749824 on Build GRCh38
rs930180367 Chr.X: 118749689-118749689 on Build GRCh38
rs749322797 Chr.X: 118749681-118749681 on Build GRCh38
rs775043802 Chr.X: 118749752-118749752 on Build GRCh38
rs368160832 Chr.X: 118749651-118749651 on Build GRCh38
rs780281252 Chr.X: 118749673-118749673 on Build GRCh38
rs756702155 Chr.X: 118749813-118749813 on Build GRCh38
rs767800605 Chr.X: 118749785-118749785 on Build GRCh38
rs778996674 Chr.X: 118749700-118749700 on Build GRCh38
rs958834724 Chr.X: 118749611-118749611 on Build GRCh38
rs746859527 Chr.X: 118749703-118749703 on Build GRCh38
rs907171658 Chr.X: 118749811-118749811 on Build GRCh38
rs368784033 Chr.X: 118749747-118749747 on Build GRCh38
rs766979631 Chr.X: 118749815-118749815 on Build GRCh38
rs755308562 Chr.X: 118749698-118749698 on Build GRCh38
rs371029411 Chr.X: 118749654-118749654 on Build GRCh38
rs776448147 Chr.X: 118749731-118749731 on Build GRCh38
rs781746124 Chr.X: 118749642-118749642 on Build GRCh38
rs762448978 Chr.X: 118749753-118749753 on Build GRCh38
rs777658944 Chr.X: 118749619-118749641 on Build GRCh38
rs201663916 Chr.X: 118749745-118749745 on Build GRCh38
rs763850965 Chr.X: 118749760-118749760 on Build GRCh38
rs757533947 Chr.X: 118749627-118749627 on Build GRCh38

View Full Product Details