Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 42163262-42163513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUCA2A
Entrez Gene ID
2980
Gene Name
guanylate cyclase activator 2A
Gene Aliases
GCAP-I, GUCA2, STARA
UniGene
Hs.778
Gene Chromosome Location
Chr.1: 42162691-42164724 on Build GRCh38
Transcripts
NM_033553.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1209196 c.223C>T p.P75S Chr.1: 42163463-42163463 on Build GRCh38
909610 c.216G>A p.P72P Chr.1: 42163470-42163470 on Build GRCh38
6889104 c.255C>T p.P85P Chr.1: 42163431-42163431 on Build GRCh38
4008029 c.233T>A p.L78H Chr.1: 42163453-42163453 on Build GRCh38
7270949 c.274C>A p.Q92K Chr.1: 42163412-42163412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2232222 Chr.1: 42163434-42163434 on Build GRCh38
rs776384590 Chr.1: 42163356-42163356 on Build GRCh38
rs796236436 Chr.1: 42163418-42163419 on Build GRCh38
rs773711464 Chr.1: 42163447-42163447 on Build GRCh38
rs377095864 Chr.1: 42163489-42163489 on Build GRCh38
rs751550135 Chr.1: 42163492-42163492 on Build GRCh38
rs145850777 Chr.1: 42163445-42163445 on Build GRCh38
rs867953441 Chr.1: 42163285-42163285 on Build GRCh38
rs1042489583 Chr.1: 42163353-42163353 on Build GRCh38
rs757872599 Chr.1: 42163404-42163404 on Build GRCh38
rs35502103 Chr.1: 42163428-42163428 on Build GRCh38
rs2232223 Chr.1: 42163376-42163376 on Build GRCh38
rs772711250 Chr.1: 42163368-42163368 on Build GRCh38
rs762798459 Chr.1: 42163436-42163436 on Build GRCh38
rs112720589 Chr.1: 42163429-42163429 on Build GRCh38
rs201815416 Chr.1: 42163478-42163478 on Build GRCh38
rs1031845364 Chr.1: 42163337-42163337 on Build GRCh38
rs777142457 Chr.1: 42163471-42163471 on Build GRCh38
rs765797714 Chr.1: 42163374-42163374 on Build GRCh38
rs759256416 Chr.1: 42163452-42163452 on Build GRCh38
rs917572734 Chr.1: 42163332-42163332 on Build GRCh38
rs754371472 Chr.1: 42163476-42163476 on Build GRCh38
rs992877035 Chr.1: 42163349-42163349 on Build GRCh38
rs751313912 Chr.1: 42163418-42163419 on Build GRCh38
rs774586138 Chr.1: 42163299-42163299 on Build GRCh38
rs752614349 Chr.1: 42163455-42163457 on Build GRCh38
rs147564899 Chr.1: 42163421-42163421 on Build GRCh38
rs767418875 Chr.1: 42163420-42163421 on Build GRCh38
rs760940245 Chr.1: 42163419-42163419 on Build GRCh38
rs759084691 Chr.1: 42163394-42163394 on Build GRCh38
rs140189038 Chr.1: 42163336-42163336 on Build GRCh38
rs956325666 Chr.1: 42163348-42163348 on Build GRCh38
rs779966689 Chr.1: 42163422-42163422 on Build GRCh38
rs770518234 Chr.1: 42163443-42163443 on Build GRCh38
rs1033518359 Chr.1: 42163481-42163481 on Build GRCh38
rs554645856 Chr.1: 42163307-42163307 on Build GRCh38
rs768708354 Chr.1: 42163427-42163427 on Build GRCh38
rs766498106 Chr.1: 42163484-42163484 on Build GRCh38
rs2232224 Chr.1: 42163369-42163369 on Build GRCh38
rs192450915 Chr.1: 42163325-42163325 on Build GRCh38
rs889390441 Chr.1: 42163378-42163378 on Build GRCh38
rs2232225 Chr.1: 42163366-42163366 on Build GRCh38
rs762451530 Chr.1: 42163382-42163382 on Build GRCh38
rs926563493 Chr.1: 42163300-42163300 on Build GRCh38
rs754207857 Chr.1: 42163401-42163401 on Build GRCh38
rs762324924 Chr.1: 42163472-42163472 on Build GRCh38
rs767239552 Chr.1: 42163470-42163470 on Build GRCh38
rs756214155 Chr.1: 42163480-42163481 on Build GRCh38
rs764569444 Chr.1: 42163397-42163397 on Build GRCh38
rs766021520 Chr.1: 42163416-42163417 on Build GRCh38
rs144332034 Chr.1: 42163425-42163425 on Build GRCh38
rs755229516 Chr.1: 42163494-42163494 on Build GRCh38
rs541903092 Chr.1: 42163479-42163479 on Build GRCh38

View Full Product Details