Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2343186-2343439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPPL2B
Entrez Gene ID
56928
Gene Name
signal peptide peptidase like 2B
Gene Aliases
IMP-4, IMP4, PSH4, PSL1
UniGene
Hs.744026
Gene Chromosome Location
Chr.19: 2328571-2355102 on Build GRCh38
Transcripts
NM_001077238.1 NM_152988.2 XM_011528138.2 XM_011528139.1 XM_011528140.1 XM_017027017.1 XM_017027018.1 XM_017027019.1

SNP Detail

SNP ID Chromosome Location
rs767853027 Chr.19: 2343299-2343299 on Build GRCh38
rs527468895 Chr.19: 2343311-2343311 on Build GRCh38
rs759770262 Chr.19: 2343295-2343295 on Build GRCh38
rs773003652 Chr.19: 2343247-2343247 on Build GRCh38
rs746660004 Chr.19: 2343220-2343220 on Build GRCh38
rs543414632 Chr.19: 2343361-2343361 on Build GRCh38
rs768355291 Chr.19: 2343227-2343227 on Build GRCh38
rs561477259 Chr.19: 2343364-2343364 on Build GRCh38
rs187824027 Chr.19: 2343254-2343254 on Build GRCh38
rs758823861 Chr.19: 2343276-2343276 on Build GRCh38
rs1016758065 Chr.19: 2343349-2343349 on Build GRCh38
rs145227878 Chr.19: 2343245-2343245 on Build GRCh38
rs746236854 Chr.19: 2343335-2343335 on Build GRCh38
rs765274596 Chr.19: 2343205-2343205 on Build GRCh38
rs779235517 Chr.19: 2343326-2343326 on Build GRCh38
rs547342033 Chr.19: 2343404-2343404 on Build GRCh38
rs955330960 Chr.19: 2343386-2343386 on Build GRCh38
rs996760389 Chr.19: 2343354-2343354 on Build GRCh38
rs545655492 Chr.19: 2343309-2343309 on Build GRCh38
rs765870732 Chr.19: 2343335-2343337 on Build GRCh38
rs574604961 Chr.19: 2343244-2343244 on Build GRCh38
rs376802313 Chr.19: 2343258-2343258 on Build GRCh38
rs747290001 Chr.19: 2343360-2343360 on Build GRCh38
rs946053435 Chr.19: 2343341-2343341 on Build GRCh38
rs913210314 Chr.19: 2343330-2343330 on Build GRCh38
rs757849317 Chr.19: 2343318-2343318 on Build GRCh38
rs191531924 Chr.19: 2343317-2343317 on Build GRCh38
rs77598010 Chr.19: 2343391-2343391 on Build GRCh38
rs895682457 Chr.19: 2343418-2343418 on Build GRCh38
rs937296055 Chr.19: 2343397-2343397 on Build GRCh38
rs1055542409 Chr.19: 2343412-2343412 on Build GRCh38
rs983696933 Chr.19: 2343234-2343234 on Build GRCh38
rs571941527 Chr.19: 2343291-2343291 on Build GRCh38
rs898433181 Chr.19: 2343352-2343352 on Build GRCh38
rs764281469 Chr.19: 2343312-2343312 on Build GRCh38
rs750570624 Chr.19: 2343275-2343275 on Build GRCh38

View Full Product Details