Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81716907-81717132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A10
Entrez Gene ID
1468
Gene Name
solute carrier family 25 member 10
Gene Aliases
DIC
UniGene
Hs.548187
Gene Chromosome Location
Chr.17: 81712236-81721016 on Build GRCh38
Transcripts
NM_001270888.1 NM_001270953.1 NM_012140.4 XM_017024220.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6342467 c.534+5G>T p.? Chr.17: 81717077-81717077 on Build GRCh38
6203378 c.479_481delTGT p.F161delF Chr.17: 81717017-81717019 on Build GRCh38
6538857 c.496A>G p.M166V Chr.17: 81717034-81717034 on Build GRCh38
7156036 c.474G>A p.R158R Chr.17: 81717012-81717012 on Build GRCh38
7156037 c.474G>A p.R158R Chr.17: 81717012-81717012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775306910 Chr.17: 81716991-81716996 on Build GRCh38
rs201512533 Chr.17: 81717054-81717054 on Build GRCh38
rs747147936 Chr.17: 81717090-81717090 on Build GRCh38
rs774614108 Chr.17: 81716974-81716975 on Build GRCh38
rs974949297 Chr.17: 81717014-81717014 on Build GRCh38
rs575068528 Chr.17: 81717007-81717007 on Build GRCh38
rs752609906 Chr.17: 81717084-81717084 on Build GRCh38
rs377325147 Chr.17: 81716981-81716981 on Build GRCh38
rs775871230 Chr.17: 81717040-81717040 on Build GRCh38
rs758430061 Chr.17: 81717001-81717001 on Build GRCh38
rs1027693548 Chr.17: 81716949-81716949 on Build GRCh38
rs370250125 Chr.17: 81717023-81717023 on Build GRCh38
rs373230757 Chr.17: 81716974-81716974 on Build GRCh38
rs777934529 Chr.17: 81717006-81717006 on Build GRCh38
rs765173372 Chr.17: 81717079-81717079 on Build GRCh38
rs764834282 Chr.17: 81717000-81717006 on Build GRCh38
rs556834379 Chr.17: 81716952-81716952 on Build GRCh38
rs1021043075 Chr.17: 81716993-81716993 on Build GRCh38
rs753746564 Chr.17: 81716984-81716984 on Build GRCh38
rs748202643 Chr.17: 81716992-81716992 on Build GRCh38
rs753624708 Chr.17: 81717051-81717051 on Build GRCh38
rs765722206 Chr.17: 81716951-81716951 on Build GRCh38
rs140229959 Chr.17: 81716967-81716968 on Build GRCh38
rs764789218 Chr.17: 81716956-81716967 on Build GRCh38
rs199532337 Chr.17: 81717046-81717046 on Build GRCh38
rs772143119 Chr.17: 81716979-81716980 on Build GRCh38
rs771258224 Chr.17: 81716970-81716971 on Build GRCh38
rs62077220 Chr.17: 81716979-81716979 on Build GRCh38
rs746284476 Chr.17: 81717024-81717024 on Build GRCh38
rs866778462 Chr.17: 81716967-81716967 on Build GRCh38
rs767135871 Chr.17: 81716964-81716964 on Build GRCh38
rs966453764 Chr.17: 81716998-81716998 on Build GRCh38
rs749908120 Chr.17: 81716955-81716956 on Build GRCh38
rs956211533 Chr.17: 81716977-81716978 on Build GRCh38
rs994906252 Chr.17: 81716930-81716930 on Build GRCh38
rs370144158 Chr.17: 81716988-81716988 on Build GRCh38
rs760596918 Chr.17: 81716991-81716992 on Build GRCh38
rs966672329 Chr.17: 81716999-81716999 on Build GRCh38
rs77612496 Chr.17: 81716978-81716979 on Build GRCh38
rs756903298 Chr.17: 81716951-81716952 on Build GRCh38
rs905683344 Chr.17: 81716929-81716929 on Build GRCh38
rs757465298 Chr.17: 81717091-81717091 on Build GRCh38
rs201109339 Chr.17: 81717050-81717050 on Build GRCh38
rs889990639 Chr.17: 81716928-81716928 on Build GRCh38
rs34675304 Chr.17: 81716967-81716967 on Build GRCh38
rs62077218 Chr.17: 81716955-81716955 on Build GRCh38
rs771191310 Chr.17: 81717016-81717016 on Build GRCh38
rs374627714 Chr.17: 81717093-81717093 on Build GRCh38
rs531566263 Chr.17: 81717085-81717085 on Build GRCh38
rs374383985 Chr.17: 81717086-81717086 on Build GRCh38
rs374616684 Chr.17: 81717035-81717035 on Build GRCh38
rs755930473 Chr.17: 81716968-81716968 on Build GRCh38

View Full Product Details