Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50448448-50448687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SBF1
Entrez Gene ID
6305
Gene Name
SET binding factor 1
Gene Aliases
CMT4B3, DENND7A, MTMR5
UniGene
Hs.589924
Gene Chromosome Location
Chr.22: 50445000-50475071 on Build GRCh38
Transcripts
NM_002972.3 XM_005261935.2 XM_011530709.2 XM_011530710.2 XM_017028905.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4582518 c.5061G>A p.R1687R Chr.22: 50448555-50448555 on Build GRCh38
4105140 c.5073T>C p.R1691R Chr.22: 50448543-50448543 on Build GRCh38
5079852 c.5073+10G>A p.? Chr.22: 50448533-50448533 on Build GRCh38
3390311 c.5073+8A>C p.? Chr.22: 50448535-50448535 on Build GRCh38
6248576 c.5001C>A p.P1667P Chr.22: 50448615-50448615 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747215974 Chr.22: 50448548-50448548 on Build GRCh38
rs777294381 Chr.22: 50448521-50448521 on Build GRCh38
rs200415227 Chr.22: 50448608-50448608 on Build GRCh38
rs188837124 Chr.22: 50448585-50448585 on Build GRCh38
rs779453976 Chr.22: 50448543-50448543 on Build GRCh38
rs775846409 Chr.22: 50448484-50448484 on Build GRCh38
rs371049216 Chr.22: 50448508-50448508 on Build GRCh38
rs755215084 Chr.22: 50448606-50448606 on Build GRCh38
rs202115500 Chr.22: 50448568-50448568 on Build GRCh38
rs541136394 Chr.22: 50448553-50448553 on Build GRCh38
rs745758511 Chr.22: 50448614-50448614 on Build GRCh38
rs753382380 Chr.22: 50448527-50448527 on Build GRCh38
rs776750233 Chr.22: 50448556-50448556 on Build GRCh38
rs368528457 Chr.22: 50448477-50448477 on Build GRCh38
rs773451056 Chr.22: 50448612-50448612 on Build GRCh38
rs762952890 Chr.22: 50448513-50448513 on Build GRCh38
rs746279946 Chr.22: 50448479-50448479 on Build GRCh38
rs780437425 Chr.22: 50448597-50448597 on Build GRCh38
rs747383843 Chr.22: 50448605-50448605 on Build GRCh38
rs914631041 Chr.22: 50448530-50448530 on Build GRCh38
rs776198448 Chr.22: 50448647-50448647 on Build GRCh38
rs760809601 Chr.22: 50448501-50448501 on Build GRCh38
rs746226600 Chr.22: 50448544-50448544 on Build GRCh38
rs990250016 Chr.22: 50448525-50448525 on Build GRCh38
rs777158689 Chr.22: 50448474-50448474 on Build GRCh38
rs146468883 Chr.22: 50448520-50448520 on Build GRCh38
rs775329310 Chr.22: 50448625-50448625 on Build GRCh38
rs768734583 Chr.22: 50448507-50448507 on Build GRCh38
rs541812137 Chr.22: 50448569-50448569 on Build GRCh38
rs531466401 Chr.22: 50448587-50448587 on Build GRCh38
rs766468828 Chr.22: 50448514-50448514 on Build GRCh38
rs760440756 Chr.22: 50448639-50448639 on Build GRCh38
rs757735364 Chr.22: 50448533-50448533 on Build GRCh38
rs762047021 Chr.22: 50448557-50448557 on Build GRCh38
rs760339634 Chr.22: 50448584-50448584 on Build GRCh38
rs367954544 Chr.22: 50448586-50448586 on Build GRCh38
rs768802405 Chr.22: 50448549-50448549 on Build GRCh38
rs772268571 Chr.22: 50448480-50448480 on Build GRCh38
rs748390126 Chr.22: 50448609-50448609 on Build GRCh38
rs775175911 Chr.22: 50448579-50448579 on Build GRCh38
rs752338849 Chr.22: 50448517-50448517 on Build GRCh38
rs554649433 Chr.22: 50448475-50448475 on Build GRCh38
rs1027321407 Chr.22: 50448645-50448645 on Build GRCh38
rs763798710 Chr.22: 50448644-50448644 on Build GRCh38
rs530167049 Chr.22: 50448564-50448564 on Build GRCh38
rs923126868 Chr.22: 50448555-50448555 on Build GRCh38
rs750453086 Chr.22: 50448545-50448545 on Build GRCh38
rs371304311 Chr.22: 50448594-50448594 on Build GRCh38
rs559398253 Chr.22: 50448563-50448563 on Build GRCh38
rs769939317 Chr.22: 50448610-50448610 on Build GRCh38
rs143914297 Chr.22: 50448516-50448516 on Build GRCh38
rs74598385 Chr.22: 50448515-50448515 on Build GRCh38
rs550066296 Chr.22: 50448615-50448615 on Build GRCh38

View Full Product Details