Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32098303-32098576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCN1
Entrez Gene ID
5954
Gene Name
reticulocalbin 1
Gene Aliases
HEL-S-84, PIG20, RCAL, RCN
UniGene
Hs.97887
Gene Chromosome Location
Chr.11: 32090931-32105726 on Build GRCh38
Transcripts
NM_002901.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5753671 c.623T>C p.V208A Chr.11: 32098524-32098524 on Build GRCh38
926355 c.509G>A p.R170H Chr.11: 32098410-32098410 on Build GRCh38
3397636 c.517A>G p.R173G Chr.11: 32098418-32098418 on Build GRCh38
428904 c.539T>A p.L180H Chr.11: 32098440-32098440 on Build GRCh38
5793491 c.513T>C p.D171D Chr.11: 32098414-32098414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756204462 Chr.11: 32098486-32098486 on Build GRCh38
rs530679508 Chr.11: 32098401-32098401 on Build GRCh38
rs751849023 Chr.11: 32098469-32098469 on Build GRCh38
rs745791740 Chr.11: 32098393-32098393 on Build GRCh38
rs760420988 Chr.11: 32098440-32098440 on Build GRCh38
rs370620780 Chr.11: 32098538-32098538 on Build GRCh38
rs117017377 Chr.11: 32098358-32098358 on Build GRCh38
rs758973071 Chr.11: 32098464-32098464 on Build GRCh38
rs753261922 Chr.11: 32098326-32098326 on Build GRCh38
rs768729169 Chr.11: 32098361-32098361 on Build GRCh38
rs781383479 Chr.11: 32098363-32098363 on Build GRCh38
rs756740384 Chr.11: 32098333-32098333 on Build GRCh38
rs763673762 Chr.11: 32098442-32098442 on Build GRCh38
rs745845082 Chr.11: 32098508-32098508 on Build GRCh38
rs780556901 Chr.11: 32098356-32098356 on Build GRCh38
rs201437677 Chr.11: 32098444-32098444 on Build GRCh38
rs1036654038 Chr.11: 32098528-32098528 on Build GRCh38
rs189574058 Chr.11: 32098475-32098475 on Build GRCh38
rs748341776 Chr.11: 32098485-32098485 on Build GRCh38
rs777825641 Chr.11: 32098493-32098493 on Build GRCh38
rs375977831 Chr.11: 32098410-32098410 on Build GRCh38
rs764785484 Chr.11: 32098459-32098459 on Build GRCh38
rs563486036 Chr.11: 32098387-32098387 on Build GRCh38
rs771919443 Chr.11: 32098409-32098409 on Build GRCh38
rs1054891734 Chr.11: 32098468-32098468 on Build GRCh38
rs766891857 Chr.11: 32098467-32098467 on Build GRCh38
rs748199752 Chr.11: 32098371-32098371 on Build GRCh38
rs746807747 Chr.11: 32098550-32098550 on Build GRCh38
rs771970472 Chr.11: 32098534-32098534 on Build GRCh38
rs769843675 Chr.11: 32098375-32098375 on Build GRCh38
rs758851383 Chr.11: 32098354-32098354 on Build GRCh38
rs373479906 Chr.11: 32098336-32098336 on Build GRCh38
rs1804282 Chr.11: 32098483-32098483 on Build GRCh38
rs751033494 Chr.11: 32098463-32098463 on Build GRCh38

View Full Product Details