Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122934669-122934902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA5
Entrez Gene ID
166378
Gene Name
spermatogenesis associated 5
Gene Aliases
AFG2, EHLMRS, SPAF
UniGene
Hs.709648
Gene Chromosome Location
Chr.4: 122923070-123319450 on Build GRCh38
Transcripts
NM_001317799.1 NM_145207.2 XM_005262783.4 XM_011531678.2 XM_011531679.2 XM_017007825.1 XM_017007826.1 XM_017007827.1 XM_017007828.1 XM_017007829.1 XM_017007830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7085285 c.1137C>G p.F379L Chr.4: 122934728-122934728 on Build GRCh38
4451193 c.1101A>T p.E367D Chr.4: 122934692-122934692 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760998804 Chr.4: 122934720-122934720 on Build GRCh38
rs1011687528 Chr.4: 122934849-122934849 on Build GRCh38
rs778659382 Chr.4: 122934692-122934692 on Build GRCh38
rs766781305 Chr.4: 122934773-122934773 on Build GRCh38
rs181978631 Chr.4: 122934805-122934805 on Build GRCh38
rs190477712 Chr.4: 122934868-122934868 on Build GRCh38
rs1056978947 Chr.4: 122934809-122934809 on Build GRCh38
rs949884350 Chr.4: 122934827-122934827 on Build GRCh38
rs537433950 Chr.4: 122934754-122934754 on Build GRCh38
rs377070684 Chr.4: 122934711-122934711 on Build GRCh38
rs553494621 Chr.4: 122934830-122934830 on Build GRCh38
rs750955601 Chr.4: 122934765-122934765 on Build GRCh38
rs919461238 Chr.4: 122934712-122934712 on Build GRCh38
rs72919901 Chr.4: 122934878-122934878 on Build GRCh38
rs187462609 Chr.4: 122934822-122934822 on Build GRCh38
rs765343136 Chr.4: 122934757-122934757 on Build GRCh38
rs139089747 Chr.4: 122934722-122934722 on Build GRCh38
rs772031133 Chr.4: 122934704-122934704 on Build GRCh38
rs1044706964 Chr.4: 122934737-122934737 on Build GRCh38
rs763421759 Chr.4: 122934770-122934770 on Build GRCh38
rs903074445 Chr.4: 122934871-122934871 on Build GRCh38
rs539688504 Chr.4: 122934825-122934825 on Build GRCh38
rs1003153079 Chr.4: 122934782-122934782 on Build GRCh38
rs776772657 Chr.4: 122934749-122934749 on Build GRCh38
rs750907309 Chr.4: 122934848-122934848 on Build GRCh38
rs375183468 Chr.4: 122934750-122934750 on Build GRCh38

View Full Product Details