Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89377596-89377869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRAGD
Entrez Gene ID
58528
Gene Name
Ras related GTP binding D
Gene Aliases
RAGD, bA11D8.2.1
UniGene
Hs.31712
Gene Chromosome Location
Chr.6: 89363509-89412470 on Build GRCh38
Transcripts
NM_021244.4 XM_005248755.4 XM_011536022.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
374905 c.902+1G>T p.? Chr.6: 89377670-89377670 on Build GRCh38
4722814 c.857G>A p.C286Y Chr.6: 89377716-89377716 on Build GRCh38
3941875 c.797T>C p.V266A Chr.6: 89377776-89377776 on Build GRCh38
3730746 c.760-10delT p.? Chr.6: 89377823-89377823 on Build GRCh38
6769758 c.830C>T p.P277L Chr.6: 89377743-89377743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951706326 Chr.6: 89377799-89377799 on Build GRCh38
rs758764437 Chr.6: 89377744-89377744 on Build GRCh38
rs749402141 Chr.6: 89377842-89377842 on Build GRCh38
rs747984622 Chr.6: 89377823-89377823 on Build GRCh38
rs550865242 Chr.6: 89377838-89377838 on Build GRCh38
rs771749834 Chr.6: 89377822-89377823 on Build GRCh38
rs768540209 Chr.6: 89377625-89377628 on Build GRCh38
rs200010421 Chr.6: 89377777-89377777 on Build GRCh38
rs1027277460 Chr.6: 89377795-89377795 on Build GRCh38
rs202246748 Chr.6: 89377822-89377822 on Build GRCh38
rs781544298 Chr.6: 89377823-89377823 on Build GRCh38
rs756214250 Chr.6: 89377832-89377832 on Build GRCh38
rs542633815 Chr.6: 89377751-89377751 on Build GRCh38
rs748439444 Chr.6: 89377830-89377830 on Build GRCh38
rs763693290 Chr.6: 89377634-89377634 on Build GRCh38
rs760488023 Chr.6: 89377628-89377628 on Build GRCh38
rs751922503 Chr.6: 89377811-89377811 on Build GRCh38
rs772055042 Chr.6: 89377623-89377623 on Build GRCh38
rs542024759 Chr.6: 89377844-89377844 on Build GRCh38
rs764706690 Chr.6: 89377740-89377740 on Build GRCh38
rs62415404 Chr.6: 89377625-89377625 on Build GRCh38
rs774240994 Chr.6: 89377628-89377628 on Build GRCh38
rs761287217 Chr.6: 89377712-89377712 on Build GRCh38
rs750936895 Chr.6: 89377742-89377742 on Build GRCh38
rs753525402 Chr.6: 89377665-89377665 on Build GRCh38

View Full Product Details