Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 101223412-101223685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNHIT1
Entrez Gene ID
10467
Gene Name
zinc finger HIT-type containing 1
Gene Aliases
CG1I, ZNFN4A1
UniGene
Hs.211079
Gene Chromosome Location
Chr.7: 101217704-101224191 on Build GRCh38
Transcripts
NM_006349.2 XM_011515739.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7093133 c.204G>T p.K68N Chr.7: 101223487-101223487 on Build GRCh38
6012829 c.194G>A p.G65E Chr.7: 101223477-101223477 on Build GRCh38
3662808 c.280A>T p.S94C Chr.7: 101223679-101223679 on Build GRCh38
1083415 c.235C>T p.R79C Chr.7: 101223518-101223518 on Build GRCh38
484529 c.254A>G p.Q85R Chr.7: 101223537-101223537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150627873 Chr.7: 101223502-101223502 on Build GRCh38
rs200176814 Chr.7: 101223637-101223637 on Build GRCh38
rs142789038 Chr.7: 101223483-101223483 on Build GRCh38
rs774215057 Chr.7: 101223457-101223457 on Build GRCh38
rs747222120 Chr.7: 101223470-101223470 on Build GRCh38
rs765885445 Chr.7: 101223508-101223508 on Build GRCh38
rs374990410 Chr.7: 101223627-101223627 on Build GRCh38
rs747895876 Chr.7: 101223489-101223489 on Build GRCh38
rs139515115 Chr.7: 101223638-101223638 on Build GRCh38
rs771865514 Chr.7: 101223632-101223632 on Build GRCh38
rs767241328 Chr.7: 101223561-101223561 on Build GRCh38
rs376064372 Chr.7: 101223585-101223585 on Build GRCh38
rs769724001 Chr.7: 101223490-101223490 on Build GRCh38
rs556415641 Chr.7: 101223622-101223622 on Build GRCh38
rs762212550 Chr.7: 101223568-101223568 on Build GRCh38
rs780206314 Chr.7: 101223468-101223468 on Build GRCh38
rs758491643 Chr.7: 101223456-101223456 on Build GRCh38
rs533890052 Chr.7: 101223498-101223498 on Build GRCh38
rs751542267 Chr.7: 101223600-101223600 on Build GRCh38
rs370797222 Chr.7: 101223598-101223598 on Build GRCh38
rs370355178 Chr.7: 101223536-101223536 on Build GRCh38
rs750936936 Chr.7: 101223571-101223571 on Build GRCh38
rs972521942 Chr.7: 101223437-101223437 on Build GRCh38
rs749229409 Chr.7: 101223624-101223624 on Build GRCh38
rs762997345 Chr.7: 101223519-101223519 on Build GRCh38
rs868171364 Chr.7: 101223493-101223494 on Build GRCh38
rs371940762 Chr.7: 101223623-101223623 on Build GRCh38
rs914427487 Chr.7: 101223577-101223577 on Build GRCh38
rs768486506 Chr.7: 101223481-101223481 on Build GRCh38
rs751639753 Chr.7: 101223663-101223663 on Build GRCh38
rs754261055 Chr.7: 101223567-101223567 on Build GRCh38
rs565536941 Chr.7: 101223655-101223655 on Build GRCh38
rs750132411 Chr.7: 101223526-101223526 on Build GRCh38
rs375361727 Chr.7: 101223602-101223602 on Build GRCh38
rs367585647 Chr.7: 101223628-101223628 on Build GRCh38
rs775268798 Chr.7: 101223636-101223636 on Build GRCh38
rs781322534 Chr.7: 101223603-101223603 on Build GRCh38
rs767058437 Chr.7: 101223494-101223495 on Build GRCh38
rs202208936 Chr.7: 101223654-101223654 on Build GRCh38
rs750939272 Chr.7: 101223644-101223644 on Build GRCh38
rs944571542 Chr.7: 101223597-101223597 on Build GRCh38
rs756889506 Chr.7: 101223488-101223495 on Build GRCh38
rs375861918 Chr.7: 101223610-101223610 on Build GRCh38
rs1051560741 Chr.7: 101223611-101223611 on Build GRCh38
rs765902856 Chr.7: 101223570-101223570 on Build GRCh38
rs990007818 Chr.7: 101223552-101223552 on Build GRCh38
rs368394346 Chr.7: 101223605-101223605 on Build GRCh38
rs773927841 Chr.7: 101223524-101223524 on Build GRCh38
rs763905018 Chr.7: 101223476-101223476 on Build GRCh38

View Full Product Details