Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 39713239-39713502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSIP1
Entrez Gene ID
161835
Gene Name
fibrous sheath interacting protein 1
Gene Aliases
HSD10
UniGene
Hs.129598
Gene Chromosome Location
Chr.15: 39598643-39782838 on Build GRCh38
Transcripts
NM_001324338.1 NM_152597.4 XM_011521305.2 XM_011521306.1 XM_011521307.2 XM_011521309.2 XM_011521311.2 XM_017021972.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5531484 c.1180G>A p.E394K Chr.15: 39713452-39713452 on Build GRCh38
7282959 c.1142G>A p.R381Q Chr.15: 39713490-39713490 on Build GRCh38
4054255 c.1188G>A p.V396V Chr.15: 39713444-39713444 on Build GRCh38
5608920 c.1188+1G>A p.? Chr.15: 39713443-39713443 on Build GRCh38
3500762 c.1141C>T p.R381W Chr.15: 39713491-39713491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371211360 Chr.15: 39713478-39713478 on Build GRCh38
rs971023851 Chr.15: 39713344-39713344 on Build GRCh38
rs757053610 Chr.15: 39713402-39713403 on Build GRCh38
rs754976627 Chr.15: 39713445-39713445 on Build GRCh38
rs557503503 Chr.15: 39713365-39713365 on Build GRCh38
rs753112409 Chr.15: 39713399-39713399 on Build GRCh38
rs778518357 Chr.15: 39713409-39713410 on Build GRCh38
rs749608294 Chr.15: 39713384-39713384 on Build GRCh38
rs1023002504 Chr.15: 39713421-39713421 on Build GRCh38
rs779660058 Chr.15: 39713442-39713443 on Build GRCh38
rs1053529582 Chr.15: 39713299-39713299 on Build GRCh38
rs751487795 Chr.15: 39713428-39713428 on Build GRCh38
rs747989942 Chr.15: 39713462-39713462 on Build GRCh38
rs7162311 Chr.15: 39713327-39713327 on Build GRCh38
rs983331004 Chr.15: 39713453-39713453 on Build GRCh38
rs958697893 Chr.15: 39713393-39713393 on Build GRCh38
rs74008680 Chr.15: 39713287-39713287 on Build GRCh38
rs893195044 Chr.15: 39713378-39713378 on Build GRCh38
rs750117823 Chr.15: 39713412-39713413 on Build GRCh38
rs537264128 Chr.15: 39713354-39713354 on Build GRCh38
rs757953034 Chr.15: 39713413-39713413 on Build GRCh38
rs749859540 Chr.15: 39713407-39713407 on Build GRCh38
rs565778799 Chr.15: 39713288-39713288 on Build GRCh38
rs889435922 Chr.15: 39713282-39713282 on Build GRCh38
rs115680773 Chr.15: 39713295-39713295 on Build GRCh38
rs377759855 Chr.15: 39713402-39713402 on Build GRCh38
rs1035616574 Chr.15: 39713355-39713355 on Build GRCh38
rs140313000 Chr.15: 39713474-39713474 on Build GRCh38
rs761310693 Chr.15: 39713401-39713401 on Build GRCh38
rs893676213 Chr.15: 39713310-39713310 on Build GRCh38
rs779586794 Chr.15: 39713413-39713413 on Build GRCh38
rs757894046 Chr.15: 39713410-39713410 on Build GRCh38
rs749514987 Chr.15: 39713475-39713475 on Build GRCh38
rs780899915 Chr.15: 39713456-39713456 on Build GRCh38
rs376642539 Chr.15: 39713321-39713321 on Build GRCh38

View Full Product Details