Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 157221480-157221753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIA2
Entrez Gene ID
2891
Gene Name
glutamate ionotropic receptor AMPA type subunit 2
Gene Aliases
GLUR2, GLURB, GluA2, GluR-K2, HBGR2
UniGene
Hs.32763
Gene Chromosome Location
Chr.4: 157205099-157370583 on Build GRCh38
Transcripts
NM_000826.3 NM_001083619.1 NM_001083620.1 XM_017008113.1 XM_017008114.1 XM_017008115.1 XM_017008116.1 XM_017008117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3129925 c.95T>C p.L32P Chr.4: 157221673-157221673 on Build GRCh38
3735663 c.100C>T p.P34S Chr.4: 157221678-157221678 on Build GRCh38
480993 c.108C>T p.G36G Chr.4: 157221686-157221686 on Build GRCh38
5128226 c.89-1delG p.? Chr.4: 157221666-157221666 on Build GRCh38
6166705 c.135A>T p.R45R Chr.4: 157221713-157221713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs909940866 Chr.4: 157221567-157221567 on Build GRCh38
rs755705423 Chr.4: 157221617-157221617 on Build GRCh38
rs868589255 Chr.4: 157221578-157221578 on Build GRCh38
rs368665384 Chr.4: 157221595-157221595 on Build GRCh38
rs749803643 Chr.4: 157221626-157221626 on Build GRCh38
rs922420366 Chr.4: 157221603-157221603 on Build GRCh38
rs201108786 Chr.4: 157221660-157221660 on Build GRCh38
rs1049626880 Chr.4: 157221616-157221616 on Build GRCh38
rs1028848693 Chr.4: 157221516-157221516 on Build GRCh38
rs112430406 Chr.4: 157221587-157221588 on Build GRCh38
rs774905233 Chr.4: 157221630-157221630 on Build GRCh38
rs200218586 Chr.4: 157221713-157221713 on Build GRCh38
rs759251598 Chr.4: 157221506-157221506 on Build GRCh38
rs755758218 Chr.4: 157221714-157221714 on Build GRCh38
rs775714722 Chr.4: 157221658-157221658 on Build GRCh38
rs140440147 Chr.4: 157221704-157221704 on Build GRCh38
rs776037451 Chr.4: 157221641-157221642 on Build GRCh38
rs28585580 Chr.4: 157221588-157221588 on Build GRCh38
rs761939874 Chr.4: 157221674-157221674 on Build GRCh38
rs967443324 Chr.4: 157221571-157221571 on Build GRCh38
rs993090327 Chr.4: 157221523-157221523 on Build GRCh38
rs367685825 Chr.4: 157221642-157221642 on Build GRCh38
rs928731593 Chr.4: 157221612-157221612 on Build GRCh38
rs916191342 Chr.4: 157221524-157221524 on Build GRCh38
rs771282129 Chr.4: 157221627-157221627 on Build GRCh38
rs551244589 Chr.4: 157221570-157221570 on Build GRCh38
rs984537355 Chr.4: 157221566-157221566 on Build GRCh38
rs532743702 Chr.4: 157221564-157221564 on Build GRCh38
rs79163474 Chr.4: 157221668-157221668 on Build GRCh38
rs1010540757 Chr.4: 157221652-157221652 on Build GRCh38
rs866122046 Chr.4: 157221722-157221722 on Build GRCh38
rs866694709 Chr.4: 157221565-157221565 on Build GRCh38
rs751482292 Chr.4: 157221687-157221687 on Build GRCh38
rs371935251 Chr.4: 157221686-157221686 on Build GRCh38
rs777347678 Chr.4: 157221727-157221727 on Build GRCh38
rs772407306 Chr.4: 157221644-157221644 on Build GRCh38
rs933857794 Chr.4: 157221605-157221605 on Build GRCh38
rs754868379 Chr.4: 157221689-157221689 on Build GRCh38
rs957529269 Chr.4: 157221559-157221559 on Build GRCh38
rs41280505 Chr.4: 157221623-157221623 on Build GRCh38

View Full Product Details