Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45397133-45397394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A30
Entrez Gene ID
253512
Gene Name
solute carrier family 25 member 30
Gene Aliases
KMCP1
UniGene
Hs.591230
Gene Chromosome Location
Chr.13: 45393316-45434016 on Build GRCh38
Transcripts
NM_001010875.3 NM_001286806.1 NM_001286807.1 XM_005266321.2 XM_006719793.3 XM_006719794.1 XM_011535026.1 XM_011535027.2 XM_017020523.1 XM_017020524.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4847024 c.762G>A p.K254K Chr.13: 45397330-45397330 on Build GRCh38
6280792 c.754-10C>T p.? Chr.13: 45397348-45397348 on Build GRCh38
3813872 c.766G>A p.E256K Chr.13: 45397326-45397326 on Build GRCh38
432427 c.801A>T p.P267P Chr.13: 45397291-45397291 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs912050834 Chr.13: 45397258-45397258 on Build GRCh38
rs371619944 Chr.13: 45397220-45397220 on Build GRCh38
rs181982887 Chr.13: 45397358-45397358 on Build GRCh38
rs950037581 Chr.13: 45397320-45397320 on Build GRCh38
rs748017114 Chr.13: 45397352-45397352 on Build GRCh38
rs781522993 Chr.13: 45397308-45397308 on Build GRCh38
rs200944205 Chr.13: 45397237-45397237 on Build GRCh38
rs767419334 Chr.13: 45397249-45397249 on Build GRCh38
rs765597494 Chr.13: 45397278-45397278 on Build GRCh38
rs944962989 Chr.13: 45397259-45397259 on Build GRCh38
rs1044079730 Chr.13: 45397321-45397321 on Build GRCh38
rs200424300 Chr.13: 45397252-45397252 on Build GRCh38
rs573986907 Chr.13: 45397352-45397352 on Build GRCh38
rs774069513 Chr.13: 45397197-45397197 on Build GRCh38
rs755728304 Chr.13: 45397334-45397334 on Build GRCh38
rs539668828 Chr.13: 45397201-45397201 on Build GRCh38
rs143559848 Chr.13: 45397307-45397307 on Build GRCh38
rs761664208 Chr.13: 45397241-45397241 on Build GRCh38
rs983651109 Chr.13: 45397224-45397224 on Build GRCh38
rs764093040 Chr.13: 45397306-45397306 on Build GRCh38
rs754868559 Chr.13: 45397314-45397315 on Build GRCh38
rs746275810 Chr.13: 45397324-45397324 on Build GRCh38
rs774288890 Chr.13: 45397239-45397239 on Build GRCh38
rs372600268 Chr.13: 45397253-45397253 on Build GRCh38
rs752894409 Chr.13: 45397295-45397295 on Build GRCh38
rs779713229 Chr.13: 45397341-45397341 on Build GRCh38
rs768080916 Chr.13: 45397238-45397238 on Build GRCh38
rs376861572 Chr.13: 45397348-45397348 on Build GRCh38
rs745515759 Chr.13: 45397212-45397212 on Build GRCh38
rs769742227 Chr.13: 45397351-45397352 on Build GRCh38
rs758456953 Chr.13: 45397300-45397300 on Build GRCh38
rs557520839 Chr.13: 45397223-45397223 on Build GRCh38
rs1016208163 Chr.13: 45397162-45397162 on Build GRCh38
rs1039299713 Chr.13: 45397280-45397280 on Build GRCh38
rs780853937 Chr.13: 45397321-45397321 on Build GRCh38
rs959377802 Chr.13: 45397235-45397235 on Build GRCh38

View Full Product Details