Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 243270048-243270305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDCCAG8
Entrez Gene ID
10806
Gene Name
serologically defined colon cancer antigen 8
Gene Aliases
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP
UniGene
Hs.591530
Gene Chromosome Location
Chr.1: 243255328-243500092 on Build GRCh38
Transcripts
NM_006642.3 XM_005273013.4 XM_005273018.1 XM_005273021.4 XM_005273022.3 XM_005273023.4 XM_006711727.3 XM_006711728.3 XM_011544021.2 XM_011544022.2 XM_011544023.2 XM_011544024.2 XM_011544026.2 XM_011544028.2 XM_011544029.2 XM_017000103.1 XM_017000104.1 XM_017000105.1 XM_017000106.1 XM_017000107.1 XM_017000108.1 XM_017000109.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4930930 c.68-2A>C p.? Chr.1: 243270103-243270103 on Build GRCh38
5053293 c.191C>T p.A64V Chr.1: 243270228-243270228 on Build GRCh38
4776696 c.131G>C p.G44A Chr.1: 243270168-243270168 on Build GRCh38
6773689 c.70C>T p.H24Y Chr.1: 243270107-243270107 on Build GRCh38
3789738 c.108G>A p.L36L Chr.1: 243270145-243270145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769624417 Chr.1: 243270198-243270198 on Build GRCh38
rs368801248 Chr.1: 243270211-243270211 on Build GRCh38
rs150953430 Chr.1: 243270136-243270136 on Build GRCh38
rs759227692 Chr.1: 243270223-243270223 on Build GRCh38
rs778405068 Chr.1: 243270154-243270154 on Build GRCh38
rs754715125 Chr.1: 243270139-243270139 on Build GRCh38
rs746020647 Chr.1: 243270174-243270174 on Build GRCh38
rs765951228 Chr.1: 243270133-243270133 on Build GRCh38
rs756518004 Chr.1: 243270236-243270236 on Build GRCh38
rs986097921 Chr.1: 243270177-243270177 on Build GRCh38
rs1037051286 Chr.1: 243270074-243270074 on Build GRCh38
rs201715098 Chr.1: 243270235-243270235 on Build GRCh38
rs776370459 Chr.1: 243270196-243270196 on Build GRCh38
rs749885268 Chr.1: 243270114-243270114 on Build GRCh38
rs779107589 Chr.1: 243270171-243270171 on Build GRCh38
rs781638035 Chr.1: 243270140-243270140 on Build GRCh38
rs35464722 Chr.1: 243270249-243270250 on Build GRCh38
rs745990713 Chr.1: 243270264-243270264 on Build GRCh38
rs759869364 Chr.1: 243270073-243270073 on Build GRCh38
rs929473088 Chr.1: 243270238-243270238 on Build GRCh38
rs772285144 Chr.1: 243270183-243270183 on Build GRCh38
rs772231725 Chr.1: 243270274-243270274 on Build GRCh38
rs200054776 Chr.1: 243270085-243270085 on Build GRCh38
rs770913941 Chr.1: 243270162-243270162 on Build GRCh38
rs778351444 Chr.1: 243270239-243270239 on Build GRCh38
rs776422382 Chr.1: 243270086-243270086 on Build GRCh38
rs149928402 Chr.1: 243270218-243270218 on Build GRCh38
rs753147253 Chr.1: 243270149-243270149 on Build GRCh38
rs370361491 Chr.1: 243270260-243270260 on Build GRCh38
rs573336252 Chr.1: 243270228-243270228 on Build GRCh38
rs554498906 Chr.1: 243270164-243270164 on Build GRCh38
rs762344881 Chr.1: 243270131-243270131 on Build GRCh38
rs200994363 Chr.1: 243270244-243270244 on Build GRCh38
rs202114636 Chr.1: 243270155-243270155 on Build GRCh38
rs186881037 Chr.1: 243270075-243270075 on Build GRCh38
rs756642876 Chr.1: 243270152-243270152 on Build GRCh38
rs761616528 Chr.1: 243270197-243270197 on Build GRCh38
rs377358593 Chr.1: 243270217-243270217 on Build GRCh38
rs757796329 Chr.1: 243270259-243270259 on Build GRCh38
rs373020926 Chr.1: 243270087-243270087 on Build GRCh38
rs1033766338 Chr.1: 243270136-243270137 on Build GRCh38

View Full Product Details