Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47449294-47449521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF41
Entrez Gene ID
7592
Gene Name
zinc finger protein 41
Gene Aliases
MRX89
UniGene
Hs.496074
Gene Chromosome Location
Chr.X: 47444691-47483671 on Build GRCh38
Transcripts
NM_001324139.1 NM_001324140.1 NM_001324141.1 NM_001324142.1 NM_001324143.1 NM_001324144.1 NM_001324145.1 NM_001324147.1 NM_001324148.1 NM_001324149.1 NM_001324150.1 NM_001324151.1 NM_001324152.1 NM_001324153.1 NM_001324154.1 NM_001324155.1 NM_001324156.1 NM_001324157.1 NM_007130.3 NM_153380.3 XM_006724550.3 XM_006724555.3 XM_017029810.1 XM_017029811.1 XM_017029812.1 XM_017029813.1 XM_017029814.1 XM_017029815.1 XM_017029816.1 XM_017029817.1

SNP Detail

SNP ID Chromosome Location
rs893979867 Chr.X: 47449480-47449480 on Build GRCh38
rs17147624 Chr.X: 47449396-47449396 on Build GRCh38
rs910399377 Chr.X: 47449331-47449331 on Build GRCh38
rs200722932 Chr.X: 47449469-47449469 on Build GRCh38
rs770145202 Chr.X: 47449371-47449371 on Build GRCh38
rs777287906 Chr.X: 47449493-47449493 on Build GRCh38
rs372311823 Chr.X: 47449330-47449330 on Build GRCh38
rs778051518 Chr.X: 47449372-47449372 on Build GRCh38
rs139056077 Chr.X: 47449346-47449346 on Build GRCh38
rs973820828 Chr.X: 47449410-47449410 on Build GRCh38
rs928226504 Chr.X: 47449465-47449465 on Build GRCh38
rs1057515903 Chr.X: 47449422-47449422 on Build GRCh38
rs865929146 Chr.X: 47449325-47449325 on Build GRCh38
rs60022654 Chr.X: 47449383-47449383 on Build GRCh38
rs867866830 Chr.X: 47449468-47449468 on Build GRCh38
rs771306630 Chr.X: 47449380-47449380 on Build GRCh38
rs765136088 Chr.X: 47449474-47449474 on Build GRCh38
rs963158974 Chr.X: 47449336-47449336 on Build GRCh38
rs937879773 Chr.X: 47449452-47449452 on Build GRCh38
rs761637546 Chr.X: 47449470-47449470 on Build GRCh38
rs375377277 Chr.X: 47449370-47449370 on Build GRCh38
rs747391741 Chr.X: 47449419-47449419 on Build GRCh38
rs746657990 Chr.X: 47449368-47449368 on Build GRCh38
rs750875263 Chr.X: 47449417-47449417 on Build GRCh38
rs104894955 Chr.X: 47449438-47449438 on Build GRCh38
rs763994798 Chr.X: 47449466-47449466 on Build GRCh38

View Full Product Details