Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48446049-48446284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRWD1
Entrez Gene ID
83743
Gene Name
glutamate rich WD repeat containing 1
Gene Aliases
CDW4, GRWD, RRB1, WDR28
UniGene
Hs.400625
Gene Chromosome Location
Chr.19: 48445773-48453907 on Build GRCh38
Transcripts
NM_031485.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1395022 c.145A>G p.M49V Chr.19: 48446150-48446150 on Build GRCh38
4651378 c.179C>T p.A60V Chr.19: 48446184-48446184 on Build GRCh38
4601507 c.98A>C p.Y33S Chr.19: 48446103-48446103 on Build GRCh38
4590627 c.109C>G p.R37G Chr.19: 48446114-48446114 on Build GRCh38
6378832 c.53A>T p.E18V Chr.19: 48446058-48446058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200635717 Chr.19: 48446105-48446105 on Build GRCh38
rs749514627 Chr.19: 48446115-48446115 on Build GRCh38
rs113956511 Chr.19: 48446195-48446195 on Build GRCh38
rs78848583 Chr.19: 48446114-48446114 on Build GRCh38
rs772590614 Chr.19: 48446147-48446147 on Build GRCh38
rs766374937 Chr.19: 48446091-48446091 on Build GRCh38
rs761578462 Chr.19: 48446084-48446084 on Build GRCh38
rs576684017 Chr.19: 48446239-48446239 on Build GRCh38
rs776279416 Chr.19: 48446073-48446073 on Build GRCh38
rs779219427 Chr.19: 48446136-48446136 on Build GRCh38
rs558393336 Chr.19: 48446130-48446130 on Build GRCh38
rs539677577 Chr.19: 48446071-48446071 on Build GRCh38
rs758376977 Chr.19: 48446088-48446088 on Build GRCh38
rs374116810 Chr.19: 48446242-48446242 on Build GRCh38
rs776202948 Chr.19: 48446198-48446198 on Build GRCh38
rs763665732 Chr.19: 48446072-48446072 on Build GRCh38
rs1034742569 Chr.19: 48446244-48446244 on Build GRCh38
rs768207196 Chr.19: 48446178-48446178 on Build GRCh38
rs750432167 Chr.19: 48446087-48446087 on Build GRCh38
rs746739721 Chr.19: 48446257-48446257 on Build GRCh38
rs762782529 Chr.19: 48446215-48446215 on Build GRCh38
rs746505181 Chr.19: 48446174-48446174 on Build GRCh38
rs370327872 Chr.19: 48446240-48446240 on Build GRCh38
rs972332571 Chr.19: 48446248-48446248 on Build GRCh38
rs1012431716 Chr.19: 48446177-48446177 on Build GRCh38
rs775917486 Chr.19: 48446163-48446163 on Build GRCh38
rs761500636 Chr.19: 48446201-48446201 on Build GRCh38
rs1001945327 Chr.19: 48446231-48446231 on Build GRCh38
rs777927195 Chr.19: 48446107-48446107 on Build GRCh38
rs775013075 Chr.19: 48446067-48446067 on Build GRCh38
rs766427847 Chr.19: 48446224-48446224 on Build GRCh38
rs752842457 Chr.19: 48446103-48446103 on Build GRCh38
rs765108642 Chr.19: 48446086-48446086 on Build GRCh38
rs1005600134 Chr.19: 48446082-48446082 on Build GRCh38
rs757613737 Chr.19: 48446135-48446135 on Build GRCh38
rs1009756394 Chr.19: 48446261-48446261 on Build GRCh38
rs771951857 Chr.19: 48446191-48446191 on Build GRCh38
rs904950831 Chr.19: 48446193-48446193 on Build GRCh38
rs919857084 Chr.19: 48446253-48446253 on Build GRCh38
rs751513065 Chr.19: 48446228-48446228 on Build GRCh38
rs765159795 Chr.19: 48446210-48446210 on Build GRCh38
rs746191977 Chr.19: 48446146-48446146 on Build GRCh38
rs111574069 Chr.19: 48446194-48446194 on Build GRCh38

View Full Product Details