Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75841167-75841418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO6
Entrez Gene ID
4646
Gene Name
myosin VI
Gene Aliases
DFNA22, DFNB37
UniGene
Hs.149387
Gene Chromosome Location
Chr.6: 75749176-75919537 on Build GRCh38
Transcripts
NM_001300899.1 NM_004999.3 XM_005248719.3 XM_005248720.3 XM_005248721.3 XM_005248722.3 XM_005248724.3 XM_005248726.3 XM_017010899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5987546 c.712C>G p.Q238E Chr.6: 75841274-75841274 on Build GRCh38
6495642 c.816+1G>A p.? Chr.6: 75841379-75841379 on Build GRCh38
5097588 c.707G>A p.C236Y Chr.6: 75841269-75841269 on Build GRCh38
6107708 c.781G>A p.E261K Chr.6: 75841343-75841343 on Build GRCh38
162750 c.793T>G p.L265V Chr.6: 75841355-75841355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766937820 Chr.6: 75841281-75841281 on Build GRCh38
rs776814111 Chr.6: 75841249-75841249 on Build GRCh38
rs121912560 Chr.6: 75841299-75841299 on Build GRCh38
rs757719710 Chr.6: 75841322-75841322 on Build GRCh38
rs759456713 Chr.6: 75841278-75841278 on Build GRCh38
rs772058658 Chr.6: 75841223-75841223 on Build GRCh38
rs755566058 Chr.6: 75841303-75841303 on Build GRCh38
rs751472354 Chr.6: 75841276-75841276 on Build GRCh38
rs775522159 Chr.6: 75841382-75841382 on Build GRCh38
rs768928382 Chr.6: 75841246-75841246 on Build GRCh38
rs746338729 Chr.6: 75841212-75841212 on Build GRCh38
rs775574293 Chr.6: 75841237-75841237 on Build GRCh38
rs777614964 Chr.6: 75841317-75841317 on Build GRCh38
rs955410532 Chr.6: 75841258-75841258 on Build GRCh38
rs772702819 Chr.6: 75841386-75841386 on Build GRCh38
rs766119690 Chr.6: 75841250-75841250 on Build GRCh38
rs760669746 Chr.6: 75841245-75841245 on Build GRCh38
rs753508452 Chr.6: 75841321-75841321 on Build GRCh38
rs746366118 Chr.6: 75841347-75841347 on Build GRCh38
rs930430931 Chr.6: 75841241-75841241 on Build GRCh38
rs768443240 Chr.6: 75841377-75841377 on Build GRCh38
rs752318101 Chr.6: 75841296-75841296 on Build GRCh38
rs1008321549 Chr.6: 75841370-75841370 on Build GRCh38
rs780471577 Chr.6: 75841205-75841206 on Build GRCh38
rs779365282 Chr.6: 75841337-75841337 on Build GRCh38
rs375645908 Chr.6: 75841207-75841207 on Build GRCh38

View Full Product Details