Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196814396-196814621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAK2
Entrez Gene ID
5062
Gene Name
p21 (RAC1) activated kinase 2
Gene Aliases
PAK65, PAKgamma
UniGene
Hs.518530
Gene Chromosome Location
Chr.3: 196739857-196832647 on Build GRCh38
Transcripts
NM_002577.4 XM_011512870.2 XM_017006501.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3380505 c.1049G>A p.R350K Chr.3: 196814564-196814564 on Build GRCh38
7105636 c.1051G>C p.E351Q Chr.3: 196814566-196814566 on Build GRCh38
6264724 c.1014G>C p.T338T Chr.3: 196814529-196814529 on Build GRCh38
1172973 c.1049G>C p.R350T Chr.3: 196814564-196814564 on Build GRCh38
7138703 c.949G>C p.D317H Chr.3: 196814464-196814464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757998972 Chr.3: 196814517-196814517 on Build GRCh38
rs927702127 Chr.3: 196814430-196814430 on Build GRCh38
rs768108246 Chr.3: 196814434-196814434 on Build GRCh38
rs376460000 Chr.3: 196814588-196814588 on Build GRCh38
rs769558175 Chr.3: 196814458-196814458 on Build GRCh38
rs772673684 Chr.3: 196814505-196814505 on Build GRCh38
rs888409864 Chr.3: 196814492-196814492 on Build GRCh38
rs776075572 Chr.3: 196814582-196814584 on Build GRCh38
rs746285007 Chr.3: 196814503-196814503 on Build GRCh38
rs758152364 Chr.3: 196814590-196814590 on Build GRCh38
rs768203249 Chr.3: 196814515-196814517 on Build GRCh38
rs369676266 Chr.3: 196814580-196814580 on Build GRCh38
rs764021697 Chr.3: 196814579-196814579 on Build GRCh38
rs373821543 Chr.3: 196814584-196814584 on Build GRCh38
rs761286680 Chr.3: 196814529-196814529 on Build GRCh38
rs534377129 Chr.3: 196814437-196814437 on Build GRCh38
rs139118165 Chr.3: 196814466-196814466 on Build GRCh38
rs746982627 Chr.3: 196814427-196814427 on Build GRCh38
rs144911835 Chr.3: 196814556-196814556 on Build GRCh38
rs774566656 Chr.3: 196814499-196814499 on Build GRCh38
rs776098582 Chr.3: 196814522-196814522 on Build GRCh38
rs189854746 Chr.3: 196814508-196814508 on Build GRCh38
rs1004209842 Chr.3: 196814559-196814559 on Build GRCh38

View Full Product Details