Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 18294153-18294398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS15
Entrez Gene ID
5651
Gene Name
transmembrane protease, serine 15
Gene Aliases
ENTK, PRSS7
UniGene
Hs.149473
Gene Chromosome Location
Chr.21: 18269107-18403806 on Build GRCh38
Transcripts
NM_002772.2 XM_011529654.1 XM_011529655.1 XM_011529656.2 XM_011529657.1 XM_011529658.1 XM_011529659.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1413444 c.2457G>A p.W819* Chr.21: 18294299-18294299 on Build GRCh38
4882129 c.2431G>A p.G811S Chr.21: 18294325-18294325 on Build GRCh38
5456843 c.2421A>G p.R807R Chr.21: 18294335-18294335 on Build GRCh38
6161124 c.2402G>A p.G801D Chr.21: 18294354-18294354 on Build GRCh38
7172262 c.2365A>G p.S789G Chr.21: 18294391-18294391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544316085 Chr.21: 18294238-18294238 on Build GRCh38
rs761304640 Chr.21: 18294359-18294359 on Build GRCh38
rs552068844 Chr.21: 18294340-18294340 on Build GRCh38
rs769957633 Chr.21: 18294326-18294326 on Build GRCh38
rs202100185 Chr.21: 18294313-18294313 on Build GRCh38
rs753574461 Chr.21: 18294269-18294269 on Build GRCh38
rs8130110 Chr.21: 18294273-18294273 on Build GRCh38
rs747421061 Chr.21: 18294275-18294275 on Build GRCh38
rs751113625 Chr.21: 18294210-18294210 on Build GRCh38
rs6517807 Chr.21: 18294209-18294209 on Build GRCh38
rs145507332 Chr.21: 18294277-18294277 on Build GRCh38
rs770587075 Chr.21: 18294288-18294288 on Build GRCh38
rs764937713 Chr.21: 18294360-18294360 on Build GRCh38
rs986609874 Chr.21: 18294329-18294329 on Build GRCh38
rs369366818 Chr.21: 18294314-18294314 on Build GRCh38
rs890500927 Chr.21: 18294207-18294207 on Build GRCh38
rs771190392 Chr.21: 18294337-18294337 on Build GRCh38
rs780890490 Chr.21: 18294361-18294361 on Build GRCh38
rs770288636 Chr.21: 18294336-18294336 on Build GRCh38
rs4395327 Chr.21: 18294213-18294213 on Build GRCh38
rs570651162 Chr.21: 18294373-18294373 on Build GRCh38
rs777935992 Chr.21: 18294332-18294332 on Build GRCh38
rs769295883 Chr.21: 18294276-18294276 on Build GRCh38
rs763270693 Chr.21: 18294287-18294287 on Build GRCh38
rs766956937 Chr.21: 18294243-18294243 on Build GRCh38
rs776218251 Chr.21: 18294357-18294357 on Build GRCh38
rs760154862 Chr.21: 18294296-18294296 on Build GRCh38
rs548456872 Chr.21: 18294289-18294289 on Build GRCh38
rs760633326 Chr.21: 18294344-18294344 on Build GRCh38
rs766240376 Chr.21: 18294225-18294225 on Build GRCh38
rs763542195 Chr.21: 18294354-18294354 on Build GRCh38
rs995868829 Chr.21: 18294321-18294321 on Build GRCh38
rs767439439 Chr.21: 18294291-18294291 on Build GRCh38
rs753392665 Chr.21: 18294311-18294311 on Build GRCh38
rs372503105 Chr.21: 18294219-18294219 on Build GRCh38
rs774505357 Chr.21: 18294339-18294339 on Build GRCh38
rs771901034 Chr.21: 18294341-18294341 on Build GRCh38
rs370207339 Chr.21: 18294325-18294325 on Build GRCh38
rs370598030 Chr.21: 18294374-18294374 on Build GRCh38
rs762636795 Chr.21: 18294224-18294224 on Build GRCh38
rs374155945 Chr.21: 18294267-18294267 on Build GRCh38
rs777389000 Chr.21: 18294263-18294263 on Build GRCh38
rs138776442 Chr.21: 18294290-18294290 on Build GRCh38
rs752210249 Chr.21: 18294253-18294253 on Build GRCh38
rs562433262 Chr.21: 18294255-18294255 on Build GRCh38
rs752686815 Chr.21: 18294294-18294294 on Build GRCh38
rs368655507 Chr.21: 18294361-18294361 on Build GRCh38
rs763633692 Chr.21: 18294300-18294300 on Build GRCh38
rs370908748 Chr.21: 18294278-18294278 on Build GRCh38
rs773278092 Chr.21: 18294284-18294284 on Build GRCh38
rs1048184002 Chr.21: 18294179-18294179 on Build GRCh38
rs975406724 Chr.21: 18294368-18294368 on Build GRCh38
rs751417137 Chr.21: 18294234-18294234 on Build GRCh38
rs372571410 Chr.21: 18294322-18294322 on Build GRCh38
rs377055842 Chr.21: 18294323-18294323 on Build GRCh38
rs775373680 Chr.21: 18294342-18294342 on Build GRCh38
rs898404041 Chr.21: 18294307-18294307 on Build GRCh38
rs768026786 Chr.21: 18294365-18294365 on Build GRCh38
rs1034438234 Chr.21: 18294299-18294299 on Build GRCh38

View Full Product Details