Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49992509-49992734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RACGAP1
Entrez Gene ID
29127
Gene Name
Rac GTPase activating protein 1
Gene Aliases
CYK4, HsCYK-4, ID-GAP, MgcRacGAP
UniGene
Hs.505469
Gene Chromosome Location
Chr.12: 49989162-50033136 on Build GRCh38
Transcripts
NM_001126103.2 NM_001126104.2 NM_001319999.1 NM_001320000.1 NM_001320001.1 NM_001320002.1 NM_001320003.1 NM_001320004.1 NM_001320005.1 NM_001320006.1 NM_001320007.1 NM_013277.4 XM_006719359.1 XM_011538238.1 XM_011538243.1 XM_017019220.1 XM_017019221.1 XM_017019222.1 XM_017019223.1 XM_017019224.1 XM_017019225.1 XM_017019226.1 XM_017019227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6000740 c.1437C>A p.H479Q Chr.12: 49992558-49992558 on Build GRCh38
2008610 c.1413C>T p.D471D Chr.12: 49992582-49992582 on Build GRCh38
1747032 c.1351G>A p.E451K Chr.12: 49992644-49992644 on Build GRCh38
6202841 c.1428C>T p.L476L Chr.12: 49992567-49992567 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777332787 Chr.12: 49992582-49992582 on Build GRCh38
rs754322726 Chr.12: 49992673-49992673 on Build GRCh38
rs771967362 Chr.12: 49992579-49992579 on Build GRCh38
rs770793317 Chr.12: 49992584-49992584 on Build GRCh38
rs761062906 Chr.12: 49992650-49992650 on Build GRCh38
rs777432882 Chr.12: 49992581-49992581 on Build GRCh38
rs753379878 Chr.12: 49992569-49992569 on Build GRCh38
rs778463915 Chr.12: 49992586-49992587 on Build GRCh38
rs750777080 Chr.12: 49992640-49992640 on Build GRCh38
rs750735816 Chr.12: 49992547-49992547 on Build GRCh38
rs763160794 Chr.12: 49992612-49992612 on Build GRCh38
rs780491460 Chr.12: 49992558-49992558 on Build GRCh38
rs778585223 Chr.12: 49992574-49992574 on Build GRCh38
rs776666448 Chr.12: 49992594-49992594 on Build GRCh38
rs768847964 Chr.12: 49992613-49992613 on Build GRCh38
rs764721491 Chr.12: 49992682-49992682 on Build GRCh38
rs761948559 Chr.12: 49992632-49992632 on Build GRCh38
rs752183253 Chr.12: 49992697-49992697 on Build GRCh38
rs766911973 Chr.12: 49992669-49992669 on Build GRCh38
rs774784118 Chr.12: 49992622-49992622 on Build GRCh38
rs756594941 Chr.12: 49992548-49992548 on Build GRCh38
rs754540026 Chr.12: 49992680-49992680 on Build GRCh38
rs757999818 Chr.12: 49992707-49992707 on Build GRCh38
rs767772123 Chr.12: 49992545-49992545 on Build GRCh38
rs554358139 Chr.12: 49992639-49992639 on Build GRCh38
rs1001853095 Chr.12: 49992663-49992663 on Build GRCh38
rs747785439 Chr.12: 49992576-49992576 on Build GRCh38

View Full Product Details