Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8862716-8862985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUC16
Entrez Gene ID
94025
Gene Name
mucin 16, cell surface associated
Gene Aliases
CA125
UniGene
Hs.432676
Gene Chromosome Location
Chr.19: 8848840-9010390 on Build GRCh38
Transcripts
NM_024690.2 XM_017027486.1 XM_017027487.1 XM_017027488.1 XM_017027489.1 XM_017027490.1 XM_017027491.1 XM_017027492.1 XM_017027493.1 XM_017027494.1 XM_017027495.1 XM_017027496.1 XM_017027497.1 XM_017027499.1 XM_017027500.1 XM_017027501.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6233536 c.42767+3A>T p.? Chr.19: 8862870-8862870 on Build GRCh38
332329 c.42762C>G p.N14254K Chr.19: 8862878-8862878 on Build GRCh38
3541789 c.42741C>T p.F14247F Chr.19: 8862899-8862899 on Build GRCh38
7222090 c.42767+10G>C p.? Chr.19: 8862863-8862863 on Build GRCh38
7419383 c.42729A>T p.L14243L Chr.19: 8862911-8862911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990593260 Chr.19: 8862756-8862756 on Build GRCh38
rs539750770 Chr.19: 8862906-8862906 on Build GRCh38
rs947765775 Chr.19: 8862738-8862738 on Build GRCh38
rs769954856 Chr.19: 8862930-8862930 on Build GRCh38
rs375713170 Chr.19: 8862886-8862886 on Build GRCh38
rs751963994 Chr.19: 8862774-8862774 on Build GRCh38
rs778738767 Chr.19: 8862899-8862899 on Build GRCh38
rs758165157 Chr.19: 8862771-8862771 on Build GRCh38
rs78005762 Chr.19: 8862786-8862786 on Build GRCh38
rs191177788 Chr.19: 8862778-8862778 on Build GRCh38
rs534387882 Chr.19: 8862856-8862856 on Build GRCh38
rs777005610 Chr.19: 8862825-8862825 on Build GRCh38
rs771415413 Chr.19: 8862955-8862955 on Build GRCh38
rs1018825167 Chr.19: 8862865-8862865 on Build GRCh38
rs374745546 Chr.19: 8862912-8862912 on Build GRCh38
rs774653604 Chr.19: 8862844-8862844 on Build GRCh38
rs749353408 Chr.19: 8862948-8862948 on Build GRCh38
rs999134879 Chr.19: 8862785-8862785 on Build GRCh38
rs372733089 Chr.19: 8862935-8862935 on Build GRCh38
rs767093694 Chr.19: 8862873-8862873 on Build GRCh38
rs774845812 Chr.19: 8862959-8862959 on Build GRCh38
rs758271332 Chr.19: 8862909-8862909 on Build GRCh38
rs370704133 Chr.19: 8862897-8862897 on Build GRCh38
rs763490488 Chr.19: 8862838-8862838 on Build GRCh38
rs200471756 Chr.19: 8862769-8862769 on Build GRCh38
rs760105792 Chr.19: 8862962-8862962 on Build GRCh38
rs928620185 Chr.19: 8862922-8862922 on Build GRCh38
rs566525814 Chr.19: 8862797-8862797 on Build GRCh38
rs773307686 Chr.19: 8862932-8862932 on Build GRCh38
rs1019726778 Chr.19: 8862892-8862892 on Build GRCh38
rs372988208 Chr.19: 8862898-8862898 on Build GRCh38
rs1053941811 Chr.19: 8862879-8862879 on Build GRCh38
rs951476764 Chr.19: 8862891-8862891 on Build GRCh38
rs769910906 Chr.19: 8862830-8862830 on Build GRCh38
rs766829810 Chr.19: 8862841-8862841 on Build GRCh38
rs748169383 Chr.19: 8862829-8862829 on Build GRCh38
rs113594656 Chr.19: 8862834-8862834 on Build GRCh38
rs752343420 Chr.19: 8862875-8862875 on Build GRCh38
rs3752137 Chr.19: 8862919-8862919 on Build GRCh38
rs76174662 Chr.19: 8862927-8862927 on Build GRCh38
rs902804884 Chr.19: 8862793-8862793 on Build GRCh38
rs61734967 Chr.19: 8862883-8862883 on Build GRCh38

View Full Product Details