Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36182260-36182531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF565
Entrez Gene ID
147929
Gene Name
zinc finger protein 565
Gene Aliases
-
UniGene
Hs.596338
Gene Chromosome Location
Chr.19: 36182060-36215084 on Build GRCh38
Transcripts
NM_001042474.2 NM_152477.4 XM_011526512.2 XM_011526514.2 XM_017026341.1 XM_017026342.1 XM_017026343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3822801 c.1474G>A p.E492K Chr.19: 36182492-36182492 on Build GRCh38
4623940 c.1493C>T p.T498I Chr.19: 36182473-36182473 on Build GRCh38
6831268 c.1442G>T p.G481V Chr.19: 36182524-36182524 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs150334930 Chr.19: 36182457-36182457 on Build GRCh38
rs776489251 Chr.19: 36182509-36182509 on Build GRCh38
rs376913280 Chr.19: 36182423-36182423 on Build GRCh38
rs892313532 Chr.19: 36182495-36182495 on Build GRCh38
rs770894365 Chr.19: 36182505-36182505 on Build GRCh38
rs543990469 Chr.19: 36182347-36182347 on Build GRCh38
rs992261138 Chr.19: 36182476-36182476 on Build GRCh38
rs1034023329 Chr.19: 36182430-36182430 on Build GRCh38
rs916122212 Chr.19: 36182383-36182383 on Build GRCh38
rs1008035541 Chr.19: 36182305-36182305 on Build GRCh38
rs16970932 Chr.19: 36182398-36182398 on Build GRCh38
rs371181899 Chr.19: 36182484-36182484 on Build GRCh38
rs779064439 Chr.19: 36182420-36182420 on Build GRCh38
rs772455496 Chr.19: 36182434-36182434 on Build GRCh38
rs968831299 Chr.19: 36182409-36182409 on Build GRCh38
rs948966762 Chr.19: 36182395-36182395 on Build GRCh38
rs1018938351 Chr.19: 36182295-36182296 on Build GRCh38
rs879735466 Chr.19: 36182408-36182408 on Build GRCh38
rs1010600812 Chr.19: 36182396-36182396 on Build GRCh38
rs377298566 Chr.19: 36182357-36182357 on Build GRCh38
rs376880678 Chr.19: 36182348-36182348 on Build GRCh38
rs1026925429 Chr.19: 36182306-36182309 on Build GRCh38
rs10425010 Chr.19: 36182296-36182296 on Build GRCh38

View Full Product Details