Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 163563551-163563824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QKI
Entrez Gene ID
9444
Gene Name
QKI, KH domain containing RNA binding
Gene Aliases
Hqk, QK, QK1, QK3, hqkI
UniGene
Hs.510324
Gene Chromosome Location
Chr.6: 163414486-163578596 on Build GRCh38
Transcripts
NM_001301085.1 NM_006775.2 NM_206853.2 NM_206854.2 NM_206855.2 XM_011536258.2 XM_011536259.2 XM_011536260.2 XM_017011502.1 XM_017011503.1 XM_017011504.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3948746 c.830C>T p.P277L Chr.6: 163563615-163563615 on Build GRCh38
420985 c.837C>T p.P279P Chr.6: 163563622-163563622 on Build GRCh38
80413 c.810A>T p.P270P Chr.6: 163563595-163563595 on Build GRCh38
4719681 c.845G>A p.G282D Chr.6: 163563630-163563630 on Build GRCh38
4586462 c.795C>T p.N265N Chr.6: 163563580-163563580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs13437446 Chr.6: 163563608-163563608 on Build GRCh38
rs548837959 Chr.6: 163563747-163563747 on Build GRCh38
rs775809461 Chr.6: 163563591-163563591 on Build GRCh38
rs760600525 Chr.6: 163563628-163563628 on Build GRCh38
rs758038769 Chr.6: 163563699-163563699 on Build GRCh38
rs138377019 Chr.6: 163563637-163563637 on Build GRCh38
rs1014914637 Chr.6: 163563730-163563730 on Build GRCh38
rs776710500 Chr.6: 163563789-163563790 on Build GRCh38
rs781764704 Chr.6: 163563734-163563734 on Build GRCh38
rs749493846 Chr.6: 163563750-163563750 on Build GRCh38
rs933689960 Chr.6: 163563601-163563601 on Build GRCh38
rs768826929 Chr.6: 163563753-163563753 on Build GRCh38
rs769837755 Chr.6: 163563580-163563580 on Build GRCh38
rs757830059 Chr.6: 163563715-163563715 on Build GRCh38
rs569768147 Chr.6: 163563622-163563622 on Build GRCh38
rs754523015 Chr.6: 163563668-163563668 on Build GRCh38
rs201095923 Chr.6: 163563771-163563771 on Build GRCh38
rs942503402 Chr.6: 163563658-163563658 on Build GRCh38
rs867595889 Chr.6: 163563588-163563588 on Build GRCh38
rs771054640 Chr.6: 163563725-163563727 on Build GRCh38
rs752041213 Chr.6: 163563712-163563712 on Build GRCh38
rs375000631 Chr.6: 163563679-163563679 on Build GRCh38
rs769937301 Chr.6: 163563741-163563741 on Build GRCh38
rs745955052 Chr.6: 163563579-163563579 on Build GRCh38
rs770788804 Chr.6: 163563792-163563792 on Build GRCh38
rs367646321 Chr.6: 163563736-163563736 on Build GRCh38
rs776582972 Chr.6: 163563795-163563795 on Build GRCh38
rs549859518 Chr.6: 163563605-163563605 on Build GRCh38
rs752418524 Chr.6: 163563698-163563698 on Build GRCh38
rs753287562 Chr.6: 163563665-163563665 on Build GRCh38
rs1006657866 Chr.6: 163563676-163563676 on Build GRCh38
rs761464048 Chr.6: 163563596-163563596 on Build GRCh38
rs778255520 Chr.6: 163563708-163563708 on Build GRCh38
rs760476034 Chr.6: 163563774-163563774 on Build GRCh38
rs371528828 Chr.6: 163563577-163563577 on Build GRCh38

View Full Product Details