Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60464177-60464450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MS4A1
Entrez Gene ID
931
Gene Name
membrane spanning 4-domains A1
Gene Aliases
B1, Bp35, CD20, CVID5, LEU-16, MS4A2, S7
UniGene
Hs.712553
Gene Chromosome Location
Chr.11: 60455809-60470752 on Build GRCh38
Transcripts
NM_021950.3 NM_152866.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6450930 c.322T>C p.S108P Chr.11: 60464330-60464330 on Build GRCh38
3397948 c.291C>T p.S97S Chr.11: 60464299-60464299 on Build GRCh38
4165862 c.280-1G>T p.? Chr.11: 60464287-60464287 on Build GRCh38
6014448 c.301C>T p.L101L Chr.11: 60464309-60464309 on Build GRCh38
1604825 c.280-10T>C p.? Chr.11: 60464278-60464278 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982624108 Chr.11: 60464381-60464381 on Build GRCh38
rs955082820 Chr.11: 60464357-60464357 on Build GRCh38
rs201138301 Chr.11: 60464257-60464257 on Build GRCh38
rs201361363 Chr.11: 60464336-60464336 on Build GRCh38
rs201472454 Chr.11: 60464391-60464391 on Build GRCh38
rs908360201 Chr.11: 60464390-60464390 on Build GRCh38
rs767956627 Chr.11: 60464269-60464269 on Build GRCh38
rs202037755 Chr.11: 60464259-60464259 on Build GRCh38
rs377012945 Chr.11: 60464362-60464362 on Build GRCh38
rs762259642 Chr.11: 60464376-60464376 on Build GRCh38
rs1022403538 Chr.11: 60464283-60464283 on Build GRCh38
rs935434584 Chr.11: 60464249-60464249 on Build GRCh38
rs200155881 Chr.11: 60464255-60464255 on Build GRCh38
rs755539451 Chr.11: 60464362-60464364 on Build GRCh38
rs374309752 Chr.11: 60464400-60464400 on Build GRCh38
rs776811532 Chr.11: 60464213-60464213 on Build GRCh38
rs768468956 Chr.11: 60464248-60464248 on Build GRCh38
rs956965652 Chr.11: 60464258-60464258 on Build GRCh38
rs751466480 Chr.11: 60464394-60464394 on Build GRCh38
rs753213417 Chr.11: 60464272-60464272 on Build GRCh38
rs369922579 Chr.11: 60464382-60464382 on Build GRCh38
rs974014134 Chr.11: 60464401-60464401 on Build GRCh38
rs762436518 Chr.11: 60464267-60464267 on Build GRCh38
rs201665706 Chr.11: 60464319-60464319 on Build GRCh38
rs374938733 Chr.11: 60464310-60464310 on Build GRCh38
rs202216356 Chr.11: 60464356-60464356 on Build GRCh38
rs775881428 Chr.11: 60464361-60464361 on Build GRCh38
rs372476401 Chr.11: 60464354-60464354 on Build GRCh38
rs376257230 Chr.11: 60464355-60464355 on Build GRCh38
rs757490182 Chr.11: 60464311-60464311 on Build GRCh38
rs763363193 Chr.11: 60464369-60464369 on Build GRCh38
rs768359203 Chr.11: 60464352-60464352 on Build GRCh38
rs201962059 Chr.11: 60464299-60464299 on Build GRCh38
rs200265912 Chr.11: 60464320-60464320 on Build GRCh38
rs770430261 Chr.11: 60464358-60464358 on Build GRCh38
rs200047477 Chr.11: 60464270-60464270 on Build GRCh38
rs201552212 Chr.11: 60464254-60464254 on Build GRCh38
rs750642845 Chr.11: 60464388-60464388 on Build GRCh38
rs764382256 Chr.11: 60464392-60464392 on Build GRCh38
rs267603055 Chr.11: 60464323-60464323 on Build GRCh38
rs546451752 Chr.11: 60464221-60464221 on Build GRCh38
rs535127803 Chr.11: 60464256-60464259 on Build GRCh38
rs200582297 Chr.11: 60464365-60464365 on Build GRCh38
rs200839432 Chr.11: 60464262-60464262 on Build GRCh38
rs201137473 Chr.11: 60464275-60464275 on Build GRCh38
rs371479947 Chr.11: 60464300-60464300 on Build GRCh38
rs200611730 Chr.11: 60464252-60464252 on Build GRCh38
rs751558749 Chr.11: 60464399-60464399 on Build GRCh38
rs754322644 Chr.11: 60464370-60464370 on Build GRCh38
rs766434836 Chr.11: 60464392-60464392 on Build GRCh38
rs200616675 Chr.11: 60464242-60464242 on Build GRCh38

View Full Product Details