Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4796100-4796373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMB6
Entrez Gene ID
5694
Gene Name
proteasome subunit beta 6
Gene Aliases
DELTA, LMPY, Y
UniGene
Hs.77060
Gene Chromosome Location
Chr.17: 4796144-4798503 on Build GRCh38
Transcripts
NM_001270481.1 NM_002798.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4642133 c.36G>A p.G12G Chr.17: 4796230-4796230 on Build GRCh38
3755632 c.63G>A p.A21A Chr.17: 4796257-4796257 on Build GRCh38
1679403 c.1A>G p.M1V Chr.17: 4796195-4796195 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778000428 Chr.17: 4796252-4796252 on Build GRCh38
rs968061810 Chr.17: 4796329-4796329 on Build GRCh38
rs766281129 Chr.17: 4796244-4796244 on Build GRCh38
rs749866643 Chr.17: 4796254-4796254 on Build GRCh38
rs976321409 Chr.17: 4796183-4796183 on Build GRCh38
rs980249539 Chr.17: 4796223-4796223 on Build GRCh38
rs775892066 Chr.17: 4796285-4796285 on Build GRCh38
rs911648398 Chr.17: 4796145-4796145 on Build GRCh38
rs780757166 Chr.17: 4796176-4796176 on Build GRCh38
rs769688987 Chr.17: 4796299-4796299 on Build GRCh38
rs1038708845 Chr.17: 4796153-4796153 on Build GRCh38
rs35891847 Chr.17: 4796223-4796223 on Build GRCh38
rs749731543 Chr.17: 4796201-4796201 on Build GRCh38
rs761713056 Chr.17: 4796220-4796220 on Build GRCh38
rs772252014 Chr.17: 4796282-4796282 on Build GRCh38
rs577817777 Chr.17: 4796351-4796351 on Build GRCh38
rs73339991 Chr.17: 4796130-4796130 on Build GRCh38
rs199655593 Chr.17: 4796310-4796310 on Build GRCh38
rs760803222 Chr.17: 4796296-4796296 on Build GRCh38
rs537197758 Chr.17: 4796161-4796161 on Build GRCh38
rs754935298 Chr.17: 4796176-4796180 on Build GRCh38
rs752324933 Chr.17: 4796250-4796250 on Build GRCh38
rs934195491 Chr.17: 4796121-4796121 on Build GRCh38
rs200484344 Chr.17: 4796208-4796208 on Build GRCh38
rs751075026 Chr.17: 4796246-4796246 on Build GRCh38
rs772893873 Chr.17: 4796301-4796301 on Build GRCh38
rs895247244 Chr.17: 4796293-4796293 on Build GRCh38
rs534238959 Chr.17: 4796337-4796337 on Build GRCh38
rs371587768 Chr.17: 4796219-4796219 on Build GRCh38
rs780845009 Chr.17: 4796249-4796249 on Build GRCh38
rs376142699 Chr.17: 4796319-4796319 on Build GRCh38
rs762784189 Chr.17: 4796242-4796242 on Build GRCh38
rs116466042 Chr.17: 4796212-4796212 on Build GRCh38
rs762740828 Chr.17: 4796306-4796306 on Build GRCh38
rs528222371 Chr.17: 4796123-4796123 on Build GRCh38
rs368206749 Chr.17: 4796324-4796324 on Build GRCh38
rs1031901285 Chr.17: 4796200-4796200 on Build GRCh38
rs550862551 Chr.17: 4796189-4796189 on Build GRCh38
rs571506548 Chr.17: 4796150-4796150 on Build GRCh38
rs938713750 Chr.17: 4796256-4796256 on Build GRCh38
rs920283823 Chr.17: 4796175-4796175 on Build GRCh38
rs943088693 Chr.17: 4796146-4796146 on Build GRCh38
rs368384009 Chr.17: 4796187-4796187 on Build GRCh38
rs775696343 Chr.17: 4796181-4796181 on Build GRCh38
rs779519710 Chr.17: 4796263-4796263 on Build GRCh38
rs754447378 Chr.17: 4796248-4796248 on Build GRCh38
rs953886710 Chr.17: 4796120-4796120 on Build GRCh38
rs956294311 Chr.17: 4796206-4796206 on Build GRCh38
rs756545316 Chr.17: 4796251-4796251 on Build GRCh38
rs750440943 Chr.17: 4796237-4796237 on Build GRCh38
rs745965445 Chr.17: 4796279-4796279 on Build GRCh38
rs751449213 Chr.17: 4796312-4796312 on Build GRCh38
rs140751461 Chr.17: 4796226-4796226 on Build GRCh38
rs764908804 Chr.17: 4796228-4796228 on Build GRCh38
rs771111047 Chr.17: 4796204-4796204 on Build GRCh38
rs3169950 Chr.17: 4796257-4796257 on Build GRCh38
rs767881546 Chr.17: 4796215-4796215 on Build GRCh38

View Full Product Details