Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8219158-8219403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3TC1
Entrez Gene ID
54436
Gene Name
SH3 domain and tetratricopeptide repeats 1
Gene Aliases
-
UniGene
Hs.479116
Gene Chromosome Location
Chr.4: 8182072-8241103 on Build GRCh38
Transcripts
NM_001318480.1 NM_018986.4 NR_134639.1 XM_006713889.2 XM_006713891.3 XM_011513488.1 XM_011513489.2 XM_017008300.1 XM_017008301.1 XM_017008302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2955368 c.980G>T p.R327L Chr.4: 8219398-8219398 on Build GRCh38
3606486 c.969G>A p.E323E Chr.4: 8219387-8219387 on Build GRCh38
4725703 c.919G>A p.V307M Chr.4: 8219337-8219337 on Build GRCh38
7094175 c.956C>T p.S319L Chr.4: 8219374-8219374 on Build GRCh38
4725704 c.979C>T p.R327* Chr.4: 8219397-8219397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929478552 Chr.4: 8219212-8219212 on Build GRCh38
rs982296010 Chr.4: 8219374-8219374 on Build GRCh38
rs918927257 Chr.4: 8219217-8219217 on Build GRCh38
rs1036761944 Chr.4: 8219182-8219182 on Build GRCh38
rs753815116 Chr.4: 8219371-8219371 on Build GRCh38
rs200686267 Chr.4: 8219340-8219340 on Build GRCh38
rs113123997 Chr.4: 8219350-8219350 on Build GRCh38
rs919392273 Chr.4: 8219199-8219199 on Build GRCh38
rs778707879 Chr.4: 8219233-8219233 on Build GRCh38
rs781769404 Chr.4: 8219330-8219330 on Build GRCh38
rs141127247 Chr.4: 8219382-8219382 on Build GRCh38
rs755101260 Chr.4: 8219293-8219293 on Build GRCh38
rs758432520 Chr.4: 8219378-8219378 on Build GRCh38
rs752736688 Chr.4: 8219377-8219377 on Build GRCh38
rs1043245067 Chr.4: 8219267-8219267 on Build GRCh38
rs1010971584 Chr.4: 8219291-8219291 on Build GRCh38
rs766142196 Chr.4: 8219366-8219366 on Build GRCh38
rs539958004 Chr.4: 8219193-8219193 on Build GRCh38
rs374311502 Chr.4: 8219317-8219317 on Build GRCh38
rs41266527 Chr.4: 8219381-8219381 on Build GRCh38
rs985347437 Chr.4: 8219332-8219332 on Build GRCh38
rs780763394 Chr.4: 8219379-8219379 on Build GRCh38
rs373764407 Chr.4: 8219351-8219351 on Build GRCh38
rs904751649 Chr.4: 8219280-8219280 on Build GRCh38
rs4696781 Chr.4: 8219269-8219269 on Build GRCh38
rs893813676 Chr.4: 8219284-8219284 on Build GRCh38
rs765418388 Chr.4: 8219375-8219375 on Build GRCh38
rs4696782 Chr.4: 8219331-8219331 on Build GRCh38
rs941970012 Chr.4: 8219200-8219200 on Build GRCh38
rs758540043 Chr.4: 8219311-8219311 on Build GRCh38
rs1002490789 Chr.4: 8219245-8219245 on Build GRCh38
rs1047026366 Chr.4: 8219228-8219228 on Build GRCh38
rs971698971 Chr.4: 8219213-8219213 on Build GRCh38
rs753033560 Chr.4: 8219298-8219298 on Build GRCh38
rs144219902 Chr.4: 8219355-8219355 on Build GRCh38
rs1055094967 Chr.4: 8219266-8219266 on Build GRCh38
rs746284969 Chr.4: 8219363-8219392 on Build GRCh38
rs529465819 Chr.4: 8219261-8219261 on Build GRCh38
rs760486101 Chr.4: 8219360-8219360 on Build GRCh38
rs761584143 Chr.4: 8219342-8219342 on Build GRCh38
rs201754307 Chr.4: 8219321-8219321 on Build GRCh38
rs572894266 Chr.4: 8219181-8219181 on Build GRCh38
rs200118219 Chr.4: 8219380-8219380 on Build GRCh38
rs550663524 Chr.4: 8219194-8219194 on Build GRCh38
rs768319393 Chr.4: 8219334-8219334 on Build GRCh38
rs933021686 Chr.4: 8219243-8219243 on Build GRCh38
rs201543520 Chr.4: 8219313-8219313 on Build GRCh38
rs772677653 Chr.4: 8219375-8219375 on Build GRCh38
rs759468100 Chr.4: 8219372-8219372 on Build GRCh38

View Full Product Details