Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45404232-45404489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A30
Entrez Gene ID
253512
Gene Name
solute carrier family 25 member 30
Gene Aliases
KMCP1
UniGene
Hs.591230
Gene Chromosome Location
Chr.13: 45393316-45434016 on Build GRCh38
Transcripts
NM_001010875.3 NM_001286806.1 NM_001286807.1 XM_005266321.2 XM_005266322.1 XM_006719793.3 XM_006719794.1 XM_011535026.1 XM_011535027.2 XM_017020523.1 XM_017020524.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5376776 c.365C>T p.T122I Chr.13: 45404355-45404355 on Build GRCh38
4726840 c.320C>T p.P107L Chr.13: 45404400-45404400 on Build GRCh38
5027765 c.354C>T p.V118V Chr.13: 45404366-45404366 on Build GRCh38
7428758 c.316_317delCT p.L106fs*19 Chr.13: 45404403-45404404 on Build GRCh38
1629112 c.351A>T p.G117G Chr.13: 45404369-45404369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757386533 Chr.13: 45404389-45404389 on Build GRCh38
rs755598475 Chr.13: 45404461-45404461 on Build GRCh38
rs756861304 Chr.13: 45404383-45404389 on Build GRCh38
rs369168176 Chr.13: 45404306-45404306 on Build GRCh38
rs371551234 Chr.13: 45404393-45404393 on Build GRCh38
rs200588795 Chr.13: 45404351-45404351 on Build GRCh38
rs764931089 Chr.13: 45404369-45404369 on Build GRCh38
rs946783279 Chr.13: 45404336-45404336 on Build GRCh38
rs781107356 Chr.13: 45404276-45404276 on Build GRCh38
rs764459489 Chr.13: 45404411-45404411 on Build GRCh38
rs759539811 Chr.13: 45404367-45404367 on Build GRCh38
rs761351687 Chr.13: 45404311-45404311 on Build GRCh38
rs774844007 Chr.13: 45404310-45404310 on Build GRCh38
rs1027389269 Chr.13: 45404264-45404264 on Build GRCh38
rs974743109 Chr.13: 45404319-45404319 on Build GRCh38
rs74481952 Chr.13: 45404444-45404444 on Build GRCh38
rs952033657 Chr.13: 45404265-45404265 on Build GRCh38
rs749825205 Chr.13: 45404304-45404304 on Build GRCh38
rs750628836 Chr.13: 45404400-45404400 on Build GRCh38
rs912108906 Chr.13: 45404302-45404302 on Build GRCh38
rs922028557 Chr.13: 45404329-45404329 on Build GRCh38
rs745996676 Chr.13: 45404279-45404279 on Build GRCh38
rs752583990 Chr.13: 45404376-45404376 on Build GRCh38
rs756205306 Chr.13: 45404422-45404422 on Build GRCh38
rs760130206 Chr.13: 45404343-45404343 on Build GRCh38
rs778295443 Chr.13: 45404438-45404439 on Build GRCh38
rs570514291 Chr.13: 45404426-45404426 on Build GRCh38
rs111271758 Chr.13: 45404280-45404280 on Build GRCh38
rs144982801 Chr.13: 45404363-45404363 on Build GRCh38
rs780659099 Chr.13: 45404296-45404296 on Build GRCh38
rs781432444 Chr.13: 45404399-45404399 on Build GRCh38
rs771797762 Chr.13: 45404314-45404314 on Build GRCh38
rs1042898222 Chr.13: 45404345-45404345 on Build GRCh38
rs772771171 Chr.13: 45404317-45404317 on Build GRCh38

View Full Product Details