Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 165090352-165090607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN3A
Entrez Gene ID
6328
Gene Name
sodium voltage-gated channel alpha subunit 3
Gene Aliases
NAC3, Nav1.3
UniGene
Hs.435274
Gene Chromosome Location
Chr.2: 165087520-165204295 on Build GRCh38
Transcripts
NM_001081676.1 NM_001081677.1 NM_006922.3 XM_011511610.2 XM_011511613.2 XM_017004660.1 XM_017004661.1 XM_017004662.1 XM_017004663.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1691247 c.5501C>T p.P1834L Chr.2: 165090505-165090505 on Build GRCh38
3568684 c.5565C>T p.A1855A Chr.2: 165090588-165090588 on Build GRCh38
4642532 c.5579G>A p.R1860H Chr.2: 165090427-165090427 on Build GRCh38
4399665 c.5532C>T p.T1844T Chr.2: 165090474-165090474 on Build GRCh38
1224858 c.5604G>T p.K1868N Chr.2: 165090402-165090402 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146618194 Chr.2: 165090570-165090570 on Build GRCh38
rs376113629 Chr.2: 165090427-165090427 on Build GRCh38
rs752441027 Chr.2: 165090561-165090561 on Build GRCh38
rs945486162 Chr.2: 165090475-165090475 on Build GRCh38
rs748414391 Chr.2: 165090474-165090474 on Build GRCh38
rs201183625 Chr.2: 165090525-165090525 on Build GRCh38
rs781506035 Chr.2: 165090464-165090464 on Build GRCh38
rs755958042 Chr.2: 165090416-165090416 on Build GRCh38
rs950595171 Chr.2: 165090457-165090457 on Build GRCh38
rs758547801 Chr.2: 165090441-165090441 on Build GRCh38
rs763004206 Chr.2: 165090434-165090434 on Build GRCh38
rs373422316 Chr.2: 165090384-165090384 on Build GRCh38
rs41265137 Chr.2: 165090569-165090569 on Build GRCh38
rs200752113 Chr.2: 165090428-165090428 on Build GRCh38
rs141161490 Chr.2: 165090495-165090495 on Build GRCh38
rs763522130 Chr.2: 165090506-165090506 on Build GRCh38
rs367631757 Chr.2: 165090440-165090440 on Build GRCh38
rs200922494 Chr.2: 165090564-165090564 on Build GRCh38
rs991413984 Chr.2: 165090515-165090515 on Build GRCh38
rs747096151 Chr.2: 165090454-165090454 on Build GRCh38
rs878854906 Chr.2: 165090497-165090497 on Build GRCh38
rs147193145 Chr.2: 165090388-165090388 on Build GRCh38
rs780334537 Chr.2: 165090442-165090442 on Build GRCh38
rs879245991 Chr.2: 165090488-165090488 on Build GRCh38
rs768691987 Chr.2: 165090463-165090463 on Build GRCh38
rs915912785 Chr.2: 165090531-165090531 on Build GRCh38
rs111957552 Chr.2: 165090456-165090456 on Build GRCh38
rs770414744 Chr.2: 165090507-165090507 on Build GRCh38
rs73969174 Chr.2: 165090483-165090483 on Build GRCh38
rs767038374 Chr.2: 165090539-165090539 on Build GRCh38
rs773793959 Chr.2: 165090517-165090517 on Build GRCh38

View Full Product Details