Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 136003339-136003612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP2R3A
Entrez Gene ID
5523
Gene Name
protein phosphatase 2 regulatory subunit B''alpha
Gene Aliases
PPP2R3, PR130, PR72
UniGene
Hs.518155
Gene Chromosome Location
Chr.3: 135965673-136147910 on Build GRCh38
Transcripts
NM_001190447.1 NM_002718.4 XM_006713686.3 XM_011512956.2 XM_017006787.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6999777 c.1982C>A p.P661Q Chr.3: 136003480-136003480 on Build GRCh38
4767132 c.1923A>C p.R641S Chr.3: 136003421-136003421 on Build GRCh38
3723171 c.1914A>T p.S638S Chr.3: 136003412-136003412 on Build GRCh38
4113825 c.1982C>T p.P661L Chr.3: 136003480-136003480 on Build GRCh38
1038817 c.1873C>A p.L625I Chr.3: 136003371-136003371 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373841502 Chr.3: 136003517-136003517 on Build GRCh38
rs758971933 Chr.3: 136003371-136003371 on Build GRCh38
rs762041486 Chr.3: 136003446-136003446 on Build GRCh38
rs139425067 Chr.3: 136003555-136003555 on Build GRCh38
rs757004858 Chr.3: 136003498-136003498 on Build GRCh38
rs950841993 Chr.3: 136003452-136003452 on Build GRCh38
rs753622644 Chr.3: 136003582-136003582 on Build GRCh38
rs17197552 Chr.3: 136003422-136003422 on Build GRCh38
rs906372050 Chr.3: 136003482-136003482 on Build GRCh38
rs747152098 Chr.3: 136003428-136003428 on Build GRCh38
rs760303227 Chr.3: 136003467-136003467 on Build GRCh38
rs772052921 Chr.3: 136003418-136003418 on Build GRCh38
rs200793795 Chr.3: 136003367-136003367 on Build GRCh38
rs763309087 Chr.3: 136003450-136003450 on Build GRCh38
rs778509910 Chr.3: 136003513-136003513 on Build GRCh38
rs768670667 Chr.3: 136003543-136003543 on Build GRCh38
rs749064509 Chr.3: 136003405-136003405 on Build GRCh38
rs928875487 Chr.3: 136003566-136003566 on Build GRCh38
rs757171592 Chr.3: 136003409-136003409 on Build GRCh38
rs763816908 Chr.3: 136003477-136003477 on Build GRCh38
rs776884129 Chr.3: 136003444-136003444 on Build GRCh38
rs1052802447 Chr.3: 136003572-136003572 on Build GRCh38
rs141649812 Chr.3: 136003411-136003411 on Build GRCh38
rs766966085 Chr.3: 136003451-136003451 on Build GRCh38
rs766868358 Chr.3: 136003375-136003375 on Build GRCh38
rs768832337 Chr.3: 136003440-136003440 on Build GRCh38
rs370938134 Chr.3: 136003447-136003447 on Build GRCh38
rs548447174 Chr.3: 136003557-136003557 on Build GRCh38
rs978317083 Chr.3: 136003395-136003395 on Build GRCh38
rs758257652 Chr.3: 136003527-136003527 on Build GRCh38
rs985350297 Chr.3: 136003454-136003454 on Build GRCh38
rs756968151 Chr.3: 136003490-136003491 on Build GRCh38
rs371828589 Chr.3: 136003567-136003567 on Build GRCh38
rs537238471 Chr.3: 136003579-136003579 on Build GRCh38
rs948199772 Chr.3: 136003448-136003448 on Build GRCh38
rs752253703 Chr.3: 136003456-136003456 on Build GRCh38
rs752262915 Chr.3: 136003387-136003387 on Build GRCh38
rs889685090 Chr.3: 136003382-136003382 on Build GRCh38
rs780077787 Chr.3: 136003541-136003541 on Build GRCh38
rs780376226 Chr.3: 136003529-136003530 on Build GRCh38
rs376280668 Chr.3: 136003542-136003542 on Build GRCh38
rs1005208744 Chr.3: 136003530-136003530 on Build GRCh38
rs773630044 Chr.3: 136003449-136003449 on Build GRCh38
rs755813305 Chr.3: 136003389-136003389 on Build GRCh38
rs753492217 Chr.3: 136003488-136003488 on Build GRCh38
rs753625274 Chr.3: 136003380-136003382 on Build GRCh38
rs938305319 Chr.3: 136003571-136003571 on Build GRCh38
rs141525835 Chr.3: 136003399-136003399 on Build GRCh38
rs572779295 Chr.3: 136003431-136003431 on Build GRCh38
rs150900402 Chr.3: 136003413-136003413 on Build GRCh38

View Full Product Details