Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16251416-16251659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO42
Entrez Gene ID
54455
Gene Name
F-box protein 42
Gene Aliases
Fbx42, JFK
UniGene
Hs.522384
Gene Chromosome Location
Chr.1: 16246840-16352491 on Build GRCh38
Transcripts
NM_018994.2 XM_006710698.3 XM_017001502.1 XM_017001503.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7101060 c.1233G>A p.L411L Chr.1: 16251591-16251591 on Build GRCh38
106391 c.1357G>T p.G453C Chr.1: 16251467-16251467 on Build GRCh38
6122400 c.1304G>A p.G435D Chr.1: 16251520-16251520 on Build GRCh38
4142975 c.1371C>T p.D457D Chr.1: 16251453-16251453 on Build GRCh38
7331219 c.1344G>A p.T448T Chr.1: 16251480-16251480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757456312 Chr.1: 16251570-16251570 on Build GRCh38
rs146081658 Chr.1: 16251467-16251467 on Build GRCh38
rs772715779 Chr.1: 16251631-16251631 on Build GRCh38
rs61731120 Chr.1: 16251639-16251639 on Build GRCh38
rs767528735 Chr.1: 16251574-16251574 on Build GRCh38
rs749167525 Chr.1: 16251593-16251593 on Build GRCh38
rs778716983 Chr.1: 16251604-16251604 on Build GRCh38
rs146643943 Chr.1: 16251609-16251609 on Build GRCh38
rs780205016 Chr.1: 16251585-16251585 on Build GRCh38
rs768563937 Chr.1: 16251523-16251523 on Build GRCh38
rs530306404 Chr.1: 16251510-16251510 on Build GRCh38
rs193223563 Chr.1: 16251620-16251620 on Build GRCh38
rs771190219 Chr.1: 16251448-16251448 on Build GRCh38
rs760070779 Chr.1: 16251553-16251553 on Build GRCh38
rs781280015 Chr.1: 16251505-16251505 on Build GRCh38
rs960274503 Chr.1: 16251532-16251532 on Build GRCh38
rs146986551 Chr.1: 16251517-16251517 on Build GRCh38
rs751803662 Chr.1: 16251480-16251480 on Build GRCh38
rs747774802 Chr.1: 16251543-16251543 on Build GRCh38
rs925986836 Chr.1: 16251568-16251568 on Build GRCh38
rs61731119 Chr.1: 16251453-16251453 on Build GRCh38
rs774147621 Chr.1: 16251530-16251530 on Build GRCh38
rs532744461 Chr.1: 16251562-16251562 on Build GRCh38
rs775964452 Chr.1: 16251561-16251561 on Build GRCh38
rs756242427 Chr.1: 16251579-16251579 on Build GRCh38
rs140717950 Chr.1: 16251446-16251446 on Build GRCh38
rs772737366 Chr.1: 16251551-16251551 on Build GRCh38
rs139727320 Chr.1: 16251594-16251594 on Build GRCh38
rs750560190 Chr.1: 16251577-16251577 on Build GRCh38
rs138827630 Chr.1: 16251512-16251512 on Build GRCh38
rs143730419 Chr.1: 16251521-16251521 on Build GRCh38
rs561367398 Chr.1: 16251481-16251481 on Build GRCh38
rs148759078 Chr.1: 16251522-16251522 on Build GRCh38
rs141160805 Chr.1: 16251554-16251554 on Build GRCh38
rs766048462 Chr.1: 16251612-16251612 on Build GRCh38

View Full Product Details