Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 108476751-108476980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC2
Entrez Gene ID
9694
Gene Name
ER membrane protein complex subunit 2
Gene Aliases
KIAA0103, TTC35
UniGene
Hs.654351
Gene Chromosome Location
Chr.8: 108443624-108486907 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5779394 c.608G>A p.G203D Chr.8: 108476798-108476798 on Build GRCh38
1169132 c.642T>G p.Y214* Chr.8: 108476832-108476832 on Build GRCh38
6653619 c.596A>C p.K199T Chr.8: 108476786-108476786 on Build GRCh38
3643906 c.622C>T p.L208F Chr.8: 108476812-108476812 on Build GRCh38
6653624 c.679A>G p.R227G Chr.8: 108476869-108476869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189315527 Chr.8: 108476812-108476812 on Build GRCh38
rs377116745 Chr.8: 108476804-108476804 on Build GRCh38
rs762189951 Chr.8: 108476792-108476792 on Build GRCh38
rs768899936 Chr.8: 108476899-108476899 on Build GRCh38
rs781153204 Chr.8: 108476861-108476861 on Build GRCh38
rs900322234 Chr.8: 108476929-108476929 on Build GRCh38
rs773760939 Chr.8: 108476923-108476923 on Build GRCh38
rs777643408 Chr.8: 108476826-108476826 on Build GRCh38
rs543443887 Chr.8: 108476919-108476919 on Build GRCh38
rs745859929 Chr.8: 108476873-108476873 on Build GRCh38
rs764386770 Chr.8: 108476785-108476785 on Build GRCh38
rs750937577 Chr.8: 108476811-108476811 on Build GRCh38
rs377099218 Chr.8: 108476890-108476890 on Build GRCh38
rs774706583 Chr.8: 108476789-108476789 on Build GRCh38
rs762219516 Chr.8: 108476907-108476907 on Build GRCh38
rs1004138497 Chr.8: 108476906-108476906 on Build GRCh38
rs1052783412 Chr.8: 108476782-108476782 on Build GRCh38
rs769904959 Chr.8: 108476887-108476887 on Build GRCh38
rs369552628 Chr.8: 108476944-108476944 on Build GRCh38
rs532498083 Chr.8: 108476947-108476947 on Build GRCh38
rs752382586 Chr.8: 108476815-108476815 on Build GRCh38
rs758136281 Chr.8: 108476823-108476823 on Build GRCh38
rs141934802 Chr.8: 108476829-108476829 on Build GRCh38
rs774653414 Chr.8: 108476900-108476900 on Build GRCh38
rs965307297 Chr.8: 108476958-108476958 on Build GRCh38
rs1002244526 Chr.8: 108476842-108476842 on Build GRCh38
rs761577621 Chr.8: 108476852-108476852 on Build GRCh38
rs761141100 Chr.8: 108476925-108476925 on Build GRCh38
rs147563717 Chr.8: 108476814-108476814 on Build GRCh38

View Full Product Details