Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 42055223-42055480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKDCC
Entrez Gene ID
91461
Gene Name
protein kinase domain containing, cytoplasmic
Gene Aliases
SGK493, Vlk
UniGene
Hs.408542
Gene Chromosome Location
Chr.2: 42048021-42058528 on Build GRCh38
Transcripts
NM_138370.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3962848 c.1128G>A p.W376* Chr.2: 42055299-42055299 on Build GRCh38
6401944 c.1120C>A p.L374I Chr.2: 42055291-42055291 on Build GRCh38
4521874 c.1135G>A p.D379N Chr.2: 42055306-42055306 on Build GRCh38
4521521 c.1115G>A p.G372E Chr.2: 42055286-42055286 on Build GRCh38
4521875 c.706G>A p.D236N Chr.2: 42055306-42055306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753344748 Chr.2: 42055407-42055407 on Build GRCh38
rs764592115 Chr.2: 42055333-42055333 on Build GRCh38
rs182948588 Chr.2: 42055452-42055452 on Build GRCh38
rs755580371 Chr.2: 42055294-42055294 on Build GRCh38
rs759723602 Chr.2: 42055311-42055311 on Build GRCh38
rs369267445 Chr.2: 42055298-42055298 on Build GRCh38
rs202171315 Chr.2: 42055398-42055398 on Build GRCh38
rs987002255 Chr.2: 42055449-42055449 on Build GRCh38
rs747912047 Chr.2: 42055358-42055358 on Build GRCh38
rs142280977 Chr.2: 42055392-42055392 on Build GRCh38
rs375308650 Chr.2: 42055281-42055281 on Build GRCh38
rs758119349 Chr.2: 42055436-42055436 on Build GRCh38
rs762191629 Chr.2: 42055396-42055396 on Build GRCh38
rs199534537 Chr.2: 42055376-42055376 on Build GRCh38
rs752278996 Chr.2: 42055434-42055434 on Build GRCh38
rs201266311 Chr.2: 42055255-42055255 on Build GRCh38
rs745968480 Chr.2: 42055389-42055389 on Build GRCh38
rs778162594 Chr.2: 42055280-42055280 on Build GRCh38
rs754056945 Chr.2: 42055245-42055245 on Build GRCh38
rs770895165 Chr.2: 42055378-42055378 on Build GRCh38
rs765473313 Chr.2: 42055312-42055312 on Build GRCh38
rs776740396 Chr.2: 42055309-42055309 on Build GRCh38
rs55771746 Chr.2: 42055353-42055353 on Build GRCh38
rs771219651 Chr.2: 42055429-42055429 on Build GRCh38
rs778723518 Chr.2: 42055356-42055356 on Build GRCh38
rs530500531 Chr.2: 42055455-42055455 on Build GRCh38
rs911497153 Chr.2: 42055268-42055268 on Build GRCh38
rs748929908 Chr.2: 42055295-42055295 on Build GRCh38
rs776650479 Chr.2: 42055384-42055384 on Build GRCh38
rs764910100 Chr.2: 42055424-42055424 on Build GRCh38
rs770985308 Chr.2: 42055301-42055301 on Build GRCh38
rs374556546 Chr.2: 42055349-42055349 on Build GRCh38
rs918786223 Chr.2: 42055243-42055243 on Build GRCh38
rs374866491 Chr.2: 42055285-42055285 on Build GRCh38
rs753432784 Chr.2: 42055354-42055354 on Build GRCh38
rs146086146 Chr.2: 42055421-42055421 on Build GRCh38
rs374897238 Chr.2: 42055377-42055377 on Build GRCh38
rs752001053 Chr.2: 42055340-42055340 on Build GRCh38
rs759811568 Chr.2: 42055247-42055247 on Build GRCh38
rs753099365 Chr.2: 42055258-42055258 on Build GRCh38
rs112427825 Chr.2: 42055394-42055394 on Build GRCh38
rs747921012 Chr.2: 42055299-42055299 on Build GRCh38
rs139834357 Chr.2: 42055300-42055300 on Build GRCh38
rs1018526522 Chr.2: 42055457-42055457 on Build GRCh38
rs879550430 Chr.2: 42055281-42055287 on Build GRCh38
rs775782223 Chr.2: 42055315-42055315 on Build GRCh38
rs200003897 Chr.2: 42055393-42055393 on Build GRCh38
rs903395363 Chr.2: 42055327-42055327 on Build GRCh38
rs1005828441 Chr.2: 42055391-42055391 on Build GRCh38
rs889378901 Chr.2: 42055348-42055348 on Build GRCh38
rs936954479 Chr.2: 42055272-42055272 on Build GRCh38
rs758897324 Chr.2: 42055277-42055277 on Build GRCh38
rs756590518 Chr.2: 42055402-42055402 on Build GRCh38

View Full Product Details