Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40902414-40902683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS18
Entrez Gene ID
57617
Gene Name
VPS18, CORVET/HOPS core subunit
Gene Aliases
PEP3
UniGene
Hs.23876
Gene Chromosome Location
Chr.15: 40894410-40903975 on Build GRCh38
Transcripts
NM_020857.2 XM_011521843.2 XM_011521844.2 XM_017022447.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6077202 c.2242G>T p.D748Y Chr.15: 40902661-40902661 on Build GRCh38
961323 c.2230C>A p.L744M Chr.15: 40902649-40902649 on Build GRCh38
7330495 c.2225C>T p.A742V Chr.15: 40902644-40902644 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1036454114 Chr.15: 40902541-40902541 on Build GRCh38
rs764969092 Chr.15: 40902566-40902566 on Build GRCh38
rs758609532 Chr.15: 40902575-40902575 on Build GRCh38
rs915569011 Chr.15: 40902495-40902495 on Build GRCh38
rs578159801 Chr.15: 40902527-40902527 on Build GRCh38
rs535207580 Chr.15: 40902473-40902473 on Build GRCh38
rs554486193 Chr.15: 40902608-40902608 on Build GRCh38
rs1045278718 Chr.15: 40902529-40902529 on Build GRCh38
rs755487795 Chr.15: 40902603-40902603 on Build GRCh38
rs1002074317 Chr.15: 40902462-40902462 on Build GRCh38
rs372625192 Chr.15: 40902633-40902633 on Build GRCh38
rs950151278 Chr.15: 40902451-40902451 on Build GRCh38
rs745572510 Chr.15: 40902656-40902656 on Build GRCh38
rs992868979 Chr.15: 40902486-40902486 on Build GRCh38
rs924329555 Chr.15: 40902477-40902478 on Build GRCh38
rs149373237 Chr.15: 40902619-40902619 on Build GRCh38
rs180978650 Chr.15: 40902485-40902485 on Build GRCh38
rs905926118 Chr.15: 40902459-40902459 on Build GRCh38
rs953915658 Chr.15: 40902618-40902618 on Build GRCh38
rs368947321 Chr.15: 40902583-40902583 on Build GRCh38
rs770201151 Chr.15: 40902625-40902625 on Build GRCh38
rs1045795000 Chr.15: 40902456-40902456 on Build GRCh38
rs147873066 Chr.15: 40902489-40902490 on Build GRCh38
rs139161590 Chr.15: 40902540-40902540 on Build GRCh38
rs370614114 Chr.15: 40902581-40902581 on Build GRCh38
rs867963225 Chr.15: 40902632-40902632 on Build GRCh38
rs893764619 Chr.15: 40902524-40902524 on Build GRCh38
rs963288446 Chr.15: 40902492-40902492 on Build GRCh38
rs909398524 Chr.15: 40902537-40902537 on Build GRCh38
rs1020638619 Chr.15: 40902528-40902528 on Build GRCh38
rs972793484 Chr.15: 40902497-40902497 on Build GRCh38
rs562943794 Chr.15: 40902600-40902600 on Build GRCh38
rs750659641 Chr.15: 40902567-40902567 on Build GRCh38
rs897129939 Chr.15: 40902447-40902447 on Build GRCh38

View Full Product Details