Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 63411853-63412126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG6
Entrez Gene ID
29929
Gene Name
ALG6, alpha-1,3-glucosyltransferase
Gene Aliases
CDG1C
UniGene
Hs.258501
Gene Chromosome Location
Chr.1: 63367590-63438562 on Build GRCh38
Transcripts
NM_013339.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6540491 c.774G>T p.Q258H Chr.1: 63412019-63412019 on Build GRCh38
5645559 c.727G>A p.V243I Chr.1: 63411972-63411972 on Build GRCh38
2238654 c.802C>T p.R268C Chr.1: 63412047-63412047 on Build GRCh38
4572239 c.683T>C p.F228S Chr.1: 63411928-63411928 on Build GRCh38
6540492 c.780G>T p.Q260H Chr.1: 63412019-63412019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767303584 Chr.1: 63411877-63411877 on Build GRCh38
rs771364886 Chr.1: 63412039-63412039 on Build GRCh38
rs769167626 Chr.1: 63412089-63412089 on Build GRCh38
rs776438636 Chr.1: 63412092-63412092 on Build GRCh38
rs755112036 Chr.1: 63411967-63411967 on Build GRCh38
rs529032571 Chr.1: 63412071-63412071 on Build GRCh38
rs61755863 Chr.1: 63411996-63411996 on Build GRCh38
rs997338018 Chr.1: 63411983-63411983 on Build GRCh38
rs752115616 Chr.1: 63412020-63412020 on Build GRCh38
rs763134964 Chr.1: 63411968-63411968 on Build GRCh38
rs769947380 Chr.1: 63411957-63411957 on Build GRCh38
rs59848367 Chr.1: 63411971-63411971 on Build GRCh38
rs185148397 Chr.1: 63411909-63411909 on Build GRCh38
rs76393703 Chr.1: 63411967-63411967 on Build GRCh38
rs886046472 Chr.1: 63411973-63411973 on Build GRCh38
rs528739746 Chr.1: 63411977-63411977 on Build GRCh38
rs747783222 Chr.1: 63411928-63411928 on Build GRCh38
rs746339839 Chr.1: 63412047-63412047 on Build GRCh38
rs765062497 Chr.1: 63412100-63412100 on Build GRCh38
rs747312707 Chr.1: 63411947-63411947 on Build GRCh38
rs771479098 Chr.1: 63411952-63411952 on Build GRCh38
rs763860669 Chr.1: 63411892-63411892 on Build GRCh38
rs374672230 Chr.1: 63411953-63411953 on Build GRCh38
rs777277219 Chr.1: 63412026-63412026 on Build GRCh38
rs754415021 Chr.1: 63411918-63411918 on Build GRCh38
rs12733241 Chr.1: 63412099-63412099 on Build GRCh38
rs763183173 Chr.1: 63411920-63411920 on Build GRCh38
rs189848560 Chr.1: 63412034-63412034 on Build GRCh38
rs754043594 Chr.1: 63412064-63412064 on Build GRCh38
rs762931650 Chr.1: 63412065-63412065 on Build GRCh38
rs771650075 Chr.1: 63411935-63411935 on Build GRCh38
rs917714377 Chr.1: 63411886-63411886 on Build GRCh38
rs752735437 Chr.1: 63412105-63412105 on Build GRCh38
rs371515667 Chr.1: 63412048-63412048 on Build GRCh38
rs751807761 Chr.1: 63411966-63411968 on Build GRCh38
rs760135479 Chr.1: 63411954-63411954 on Build GRCh38
rs766307079 Chr.1: 63411955-63411957 on Build GRCh38
rs376678717 Chr.1: 63412097-63412097 on Build GRCh38
rs755990716 Chr.1: 63411896-63411896 on Build GRCh38
rs12733227 Chr.1: 63412023-63412023 on Build GRCh38
rs774373539 Chr.1: 63412077-63412077 on Build GRCh38
rs367674567 Chr.1: 63411972-63411972 on Build GRCh38
rs112310201 Chr.1: 63411976-63411976 on Build GRCh38
rs776117879 Chr.1: 63412055-63412055 on Build GRCh38
rs766109595 Chr.1: 63412091-63412091 on Build GRCh38
rs144939122 Chr.1: 63412040-63412040 on Build GRCh38
rs767943869 Chr.1: 63412032-63412033 on Build GRCh38
rs886548508 Chr.1: 63412025-63412027 on Build GRCh38
rs778405436 Chr.1: 63411921-63411921 on Build GRCh38
rs945007225 Chr.1: 63411899-63411899 on Build GRCh38
rs777520437 Chr.1: 63411938-63411938 on Build GRCh38

View Full Product Details