Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 78109359-78109863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG8
Entrez Gene ID
79053
Gene Name
ALG8, alpha-1,3-glucosyltransferase
Gene Aliases
CDG1H
UniGene
Hs.503368
Gene Chromosome Location
Chr.11: 78100942-78139660 on Build GRCh38
Transcripts
NM_001007027.2 NM_024079.4 XM_005274247.3 XM_011545252.2 XM_017018274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2019232 c.952A>G p.S318G Chr.11: 78109528-78109528 on Build GRCh38
6885072 c.944C>T p.S315L Chr.11: 78109536-78109536 on Build GRCh38
429910 c.988C>T p.P330S Chr.11: 78109492-78109492 on Build GRCh38
932240 c.957T>C p.G319G Chr.11: 78109523-78109523 on Build GRCh38
6317076 c.934C>A p.P312T Chr.11: 78109546-78109546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969288651 Chr.11: 78109798-78109798 on Build GRCh38
rs901605983 Chr.11: 78109527-78109527 on Build GRCh38
rs979373947 Chr.11: 78109814-78109814 on Build GRCh38
rs1057507922 Chr.11: 78109833-78109833 on Build GRCh38
rs753052197 Chr.11: 78109480-78109480 on Build GRCh38
rs767311729 Chr.11: 78109519-78109519 on Build GRCh38
rs569975312 Chr.11: 78109802-78109802 on Build GRCh38
rs150213168 Chr.11: 78109558-78109558 on Build GRCh38
rs749148249 Chr.11: 78109545-78109545 on Build GRCh38
rs1789032 Chr.11: 78109627-78109627 on Build GRCh38
rs760264923 Chr.11: 78109596-78109596 on Build GRCh38
rs763981094 Chr.11: 78109435-78109435 on Build GRCh38
rs589272 Chr.11: 78109603-78109603 on Build GRCh38
rs532950586 Chr.11: 78109726-78109726 on Build GRCh38
rs377427118 Chr.11: 78109631-78109631 on Build GRCh38
rs749602836 Chr.11: 78109787-78109787 on Build GRCh38
rs373303136 Chr.11: 78109490-78109490 on Build GRCh38
rs868202602 Chr.11: 78109539-78109539 on Build GRCh38
rs772717043 Chr.11: 78109594-78109594 on Build GRCh38
rs763459268 Chr.11: 78109628-78109628 on Build GRCh38
rs771577945 Chr.11: 78109593-78109593 on Build GRCh38
rs898688314 Chr.11: 78109641-78109641 on Build GRCh38
rs761366888 Chr.11: 78109446-78109446 on Build GRCh38
rs749133314 Chr.11: 78109596-78109600 on Build GRCh38
rs116370749 Chr.11: 78109702-78109702 on Build GRCh38
rs186150624 Chr.11: 78109405-78109405 on Build GRCh38
rs777040526 Chr.11: 78109404-78109404 on Build GRCh38
rs141068538 Chr.11: 78109500-78109500 on Build GRCh38
rs975240498 Chr.11: 78109730-78109730 on Build GRCh38
rs145198241 Chr.11: 78109749-78109749 on Build GRCh38
rs1783515 Chr.11: 78109624-78109624 on Build GRCh38
rs781673169 Chr.11: 78109776-78109776 on Build GRCh38
rs202112771 Chr.11: 78109441-78109441 on Build GRCh38
rs912454876 Chr.11: 78109701-78109701 on Build GRCh38
rs746092328 Chr.11: 78109520-78109520 on Build GRCh38
rs1025700798 Chr.11: 78109732-78109732 on Build GRCh38
rs770011112 Chr.11: 78109525-78109525 on Build GRCh38
rs500746 Chr.11: 78109598-78109598 on Build GRCh38
rs757467776 Chr.11: 78109496-78109496 on Build GRCh38
rs988484755 Chr.11: 78109694-78109694 on Build GRCh38
rs1050059096 Chr.11: 78109619-78109619 on Build GRCh38
rs779744393 Chr.11: 78109535-78109535 on Build GRCh38
rs479415 Chr.11: 78109716-78109716 on Build GRCh38
rs191655562 Chr.11: 78109471-78109471 on Build GRCh38
rs761737089 Chr.11: 78109577-78109577 on Build GRCh38
rs755722559 Chr.11: 78109468-78109468 on Build GRCh38
rs768420996 Chr.11: 78109552-78109552 on Build GRCh38
rs770980439 Chr.11: 78109598-78109600 on Build GRCh38
rs767003895 Chr.11: 78109457-78109457 on Build GRCh38
rs747842029 Chr.11: 78109788-78109788 on Build GRCh38
rs544531 Chr.11: 78109816-78109816 on Build GRCh38
rs916469574 Chr.11: 78109427-78109427 on Build GRCh38
rs371928572 Chr.11: 78109413-78109413 on Build GRCh38
rs925139349 Chr.11: 78109469-78109469 on Build GRCh38
rs376371373 Chr.11: 78109425-78109425 on Build GRCh38
rs752380348 Chr.11: 78109572-78109572 on Build GRCh38
rs758856513 Chr.11: 78109487-78109487 on Build GRCh38
rs1052213134 Chr.11: 78109512-78109512 on Build GRCh38
rs775300260 Chr.11: 78109421-78109421 on Build GRCh38
rs764538338 Chr.11: 78109630-78109630 on Build GRCh38
rs777686455 Chr.11: 78109498-78109499 on Build GRCh38
rs769483110 Chr.11: 78109408-78109408 on Build GRCh38
rs1023683003 Chr.11: 78109763-78109763 on Build GRCh38
rs925091495 Chr.11: 78109817-78109817 on Build GRCh38
rs1013550837 Chr.11: 78109750-78109750 on Build GRCh38
rs480294 Chr.11: 78109796-78109796 on Build GRCh38
rs147646844 Chr.11: 78109832-78109832 on Build GRCh38
rs369578883 Chr.11: 78109463-78109463 on Build GRCh38

View Full Product Details