Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111908287-111908818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYAB
Entrez Gene ID
1410
Gene Name
crystallin alpha B
Gene Aliases
CMD1II, CRYA2, CTPP2, CTRCT16, HEL-S-101, HSPB5, MFM2
UniGene
Hs.53454
Gene Chromosome Location
Chr.11: 111908620-111913213 on Build GRCh38
Transcripts
NM_001289807.1 NM_001289808.1 NM_001885.2 XM_011542608.1 XM_011542609.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1351251 c.511G>A p.A171T Chr.11: 111908781-111908781 on Build GRCh38
7306583 c.488G>A p.R163H Chr.11: 111908804-111908804 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782573208 Chr.11: 111908669-111908669 on Build GRCh38
rs538657643 Chr.11: 111908570-111908570 on Build GRCh38
rs567120454 Chr.11: 111908363-111908363 on Build GRCh38
rs147970333 Chr.11: 111908782-111908782 on Build GRCh38
rs370803064 Chr.11: 111908781-111908781 on Build GRCh38
rs555182048 Chr.11: 111908362-111908362 on Build GRCh38
rs782028221 Chr.11: 111908758-111908758 on Build GRCh38
rs782808506 Chr.11: 111908725-111908725 on Build GRCh38
rs975256836 Chr.11: 111908668-111908668 on Build GRCh38
rs375893025 Chr.11: 111908761-111908761 on Build GRCh38
rs1022643136 Chr.11: 111908636-111908636 on Build GRCh38
rs941038670 Chr.11: 111908744-111908744 on Build GRCh38
rs577636561 Chr.11: 111908509-111908509 on Build GRCh38
rs374314000 Chr.11: 111908317-111908317 on Build GRCh38
rs782723002 Chr.11: 111908729-111908729 on Build GRCh38
rs782446124 Chr.11: 111908737-111908738 on Build GRCh38
rs957581668 Chr.11: 111908572-111908572 on Build GRCh38
rs963892207 Chr.11: 111908654-111908654 on Build GRCh38
rs3179808 Chr.11: 111908624-111908624 on Build GRCh38
rs1031700722 Chr.11: 111908560-111908560 on Build GRCh38
rs1045891233 Chr.11: 111908527-111908527 on Build GRCh38
rs908265346 Chr.11: 111908737-111908737 on Build GRCh38
rs1801966 Chr.11: 111908766-111908766 on Build GRCh38
rs573621530 Chr.11: 111908618-111908618 on Build GRCh38
rs781923307 Chr.11: 111908797-111908797 on Build GRCh38
rs112204677 Chr.11: 111908628-111908628 on Build GRCh38
rs370347325 Chr.11: 111908748-111908748 on Build GRCh38
rs782149494 Chr.11: 111908718-111908718 on Build GRCh38
rs955156505 Chr.11: 111908696-111908696 on Build GRCh38
rs782361571 Chr.11: 111908788-111908788 on Build GRCh38
rs907323445 Chr.11: 111908534-111908534 on Build GRCh38
rs782665062 Chr.11: 111908453-111908453 on Build GRCh38
rs562325060 Chr.11: 111908713-111908713 on Build GRCh38
rs180890418 Chr.11: 111908508-111908508 on Build GRCh38
rs558296591 Chr.11: 111908551-111908551 on Build GRCh38
rs1011552667 Chr.11: 111908576-111908576 on Build GRCh38
rs374078061 Chr.11: 111908760-111908760 on Build GRCh38
rs540926728 Chr.11: 111908704-111908704 on Build GRCh38
rs782435315 Chr.11: 111908751-111908751 on Build GRCh38
rs781853968 Chr.11: 111908726-111908726 on Build GRCh38
rs143500486 Chr.11: 111908776-111908776 on Build GRCh38
rs886047686 Chr.11: 111908657-111908657 on Build GRCh38
rs782491284 Chr.11: 111908614-111908614 on Build GRCh38
rs11549441 Chr.11: 111908778-111908778 on Build GRCh38
rs537595691 Chr.11: 111908378-111908378 on Build GRCh38
rs148052026 Chr.11: 111908610-111908610 on Build GRCh38

View Full Product Details