Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1212463-1212958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF4
Entrez Gene ID
7293
Gene Name
TNF receptor superfamily member 4
Gene Aliases
ACT35, CD134, IMD16, OX40, TXGP1L
UniGene
Hs.129780
Gene Chromosome Location
Chr.1: 1211326-1216812 on Build GRCh38
Transcripts
NM_003327.3 XM_011542074.2 XM_011542075.2 XM_011542076.2 XM_011542077.2 XM_017002231.1 XM_017002232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6345230 c.408C>A p.G136G Chr.1: 1212667-1212667 on Build GRCh38
3471568 c.381C>T p.P127P Chr.1: 1212694-1212694 on Build GRCh38
4923082 c.422G>T p.C141F Chr.1: 1212653-1212653 on Build GRCh38
4491464 c.380C>T p.P127L Chr.1: 1212695-1212695 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1014287868 Chr.1: 1212864-1212864 on Build GRCh38
rs773544484 Chr.1: 1212596-1212597 on Build GRCh38
rs965184741 Chr.1: 1212590-1212590 on Build GRCh38
rs34108055 Chr.1: 1212720-1212720 on Build GRCh38
rs568454867 Chr.1: 1212532-1212532 on Build GRCh38
rs1015997962 Chr.1: 1212872-1212872 on Build GRCh38
rs1045322678 Chr.1: 1212809-1212809 on Build GRCh38
rs963164155 Chr.1: 1212878-1212878 on Build GRCh38
rs760909412 Chr.1: 1212597-1212597 on Build GRCh38
rs919968398 Chr.1: 1212603-1212604 on Build GRCh38
rs566351921 Chr.1: 1212630-1212630 on Build GRCh38
rs968192281 Chr.1: 1212520-1212520 on Build GRCh38
rs370839049 Chr.1: 1212925-1212925 on Build GRCh38
rs1000965986 Chr.1: 1212545-1212545 on Build GRCh38
rs886166221 Chr.1: 1212850-1212850 on Build GRCh38
rs1009308690 Chr.1: 1212489-1212489 on Build GRCh38
rs367869917 Chr.1: 1212667-1212667 on Build GRCh38
rs756014065 Chr.1: 1212602-1212602 on Build GRCh38
rs796887398 Chr.1: 1212913-1212913 on Build GRCh38
rs376366886 Chr.1: 1212725-1212725 on Build GRCh38
rs747493636 Chr.1: 1212681-1212681 on Build GRCh38
rs776699361 Chr.1: 1212597-1212598 on Build GRCh38
rs538592266 Chr.1: 1212697-1212697 on Build GRCh38
rs972704269 Chr.1: 1212899-1212899 on Build GRCh38
rs766198736 Chr.1: 1212746-1212746 on Build GRCh38
rs538075572 Chr.1: 1212766-1212766 on Build GRCh38
rs534218683 Chr.1: 1212599-1212599 on Build GRCh38
rs948230090 Chr.1: 1212778-1212778 on Build GRCh38
rs11260559 Chr.1: 1212603-1212603 on Build GRCh38
rs771531096 Chr.1: 1212652-1212652 on Build GRCh38
rs768538505 Chr.1: 1212620-1212620 on Build GRCh38
rs976881951 Chr.1: 1212555-1212572 on Build GRCh38
rs867262153 Chr.1: 1212918-1212918 on Build GRCh38
rs950528702 Chr.1: 1212485-1212485 on Build GRCh38
rs868275021 Chr.1: 1212657-1212657 on Build GRCh38
rs1018049648 Chr.1: 1212534-1212534 on Build GRCh38
rs959920454 Chr.1: 1212598-1212598 on Build GRCh38
rs34588780 Chr.1: 1212898-1212898 on Build GRCh38
rs1000558686 Chr.1: 1212808-1212808 on Build GRCh38
rs554796749 Chr.1: 1212827-1212827 on Build GRCh38
rs1020629942 Chr.1: 1212494-1212494 on Build GRCh38
rs112862217 Chr.1: 1212617-1212617 on Build GRCh38
rs34082512 Chr.1: 1212499-1212499 on Build GRCh38
rs577184131 Chr.1: 1212926-1212926 on Build GRCh38
rs866150046 Chr.1: 1212660-1212660 on Build GRCh38
rs987909005 Chr.1: 1212690-1212690 on Build GRCh38
rs954634143 Chr.1: 1212922-1212922 on Build GRCh38
rs753866343 Chr.1: 1212747-1212747 on Build GRCh38
rs33986159 Chr.1: 1212605-1212606 on Build GRCh38
rs369938262 Chr.1: 1212748-1212748 on Build GRCh38
rs752647061 Chr.1: 1212600-1212601 on Build GRCh38
rs778519697 Chr.1: 1212624-1212624 on Build GRCh38
rs1057204342 Chr.1: 1212828-1212828 on Build GRCh38
rs764207404 Chr.1: 1212726-1212726 on Build GRCh38
rs772466274 Chr.1: 1212591-1212601 on Build GRCh38
rs961394549 Chr.1: 1212601-1212601 on Build GRCh38
rs974620042 Chr.1: 1212650-1212650 on Build GRCh38
rs771330950 Chr.1: 1212684-1212684 on Build GRCh38
rs34411844 Chr.1: 1212577-1212578 on Build GRCh38
rs867826495 Chr.1: 1212924-1212924 on Build GRCh38
rs983265247 Chr.1: 1212486-1212486 on Build GRCh38
rs750304065 Chr.1: 1212600-1212600 on Build GRCh38
rs959564399 Chr.1: 1212832-1212832 on Build GRCh38
rs920020627 Chr.1: 1212905-1212905 on Build GRCh38
rs1005055301 Chr.1: 1212863-1212863 on Build GRCh38
rs540161798 Chr.1: 1212849-1212849 on Build GRCh38
rs11260560 Chr.1: 1212605-1212605 on Build GRCh38
rs906819513 Chr.1: 1212817-1212817 on Build GRCh38
rs867377314 Chr.1: 1212658-1212658 on Build GRCh38
rs759479695 Chr.1: 1212598-1212599 on Build GRCh38
rs757421593 Chr.1: 1212732-1212732 on Build GRCh38
rs574834791 Chr.1: 1212843-1212843 on Build GRCh38
rs765349318 Chr.1: 1212599-1212600 on Build GRCh38

View Full Product Details