Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4915631-4916148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNA1
Entrez Gene ID
3736
Gene Name
potassium voltage-gated channel subfamily A member 1
Gene Aliases
AEMK, EA1, HBK1, HUK1, KV1.1, MBK1, MK1, RBK1
UniGene
Hs.416139
Gene Chromosome Location
Chr.12: 4909907-4918256 on Build GRCh38
Transcripts
NM_000217.2

SNP Detail

SNP ID Chromosome Location
rs777765225 Chr.12: 4916091-4916091 on Build GRCh38
rs965735741 Chr.12: 4915894-4915894 on Build GRCh38
rs1056610821 Chr.12: 4915750-4915750 on Build GRCh38
rs1049329326 Chr.12: 4915814-4915814 on Build GRCh38
rs925546420 Chr.12: 4915860-4915860 on Build GRCh38
rs758456685 Chr.12: 4916029-4916029 on Build GRCh38
rs990459977 Chr.12: 4915946-4915946 on Build GRCh38
rs765376770 Chr.12: 4915871-4915871 on Build GRCh38
rs1042477620 Chr.12: 4915695-4915695 on Build GRCh38
rs1045118207 Chr.12: 4915913-4915913 on Build GRCh38
rs922015264 Chr.12: 4916071-4916071 on Build GRCh38
rs886049527 Chr.12: 4916066-4916066 on Build GRCh38
rs112092813 Chr.12: 4915917-4915917 on Build GRCh38
rs878938800 Chr.12: 4916040-4916040 on Build GRCh38
rs866462593 Chr.12: 4915850-4915850 on Build GRCh38
rs895318457 Chr.12: 4915773-4915773 on Build GRCh38
rs753755008 Chr.12: 4915739-4915739 on Build GRCh38
rs561344869 Chr.12: 4915877-4915877 on Build GRCh38
rs946767213 Chr.12: 4915689-4915689 on Build GRCh38
rs763945925 Chr.12: 4915716-4915716 on Build GRCh38
rs948274128 Chr.12: 4916102-4916102 on Build GRCh38
rs990102407 Chr.12: 4915680-4915680 on Build GRCh38
rs542921545 Chr.12: 4915676-4915676 on Build GRCh38
rs1004546607 Chr.12: 4915852-4915852 on Build GRCh38
rs1052647284 Chr.12: 4915875-4915875 on Build GRCh38
rs181233186 Chr.12: 4916042-4916042 on Build GRCh38
rs562792530 Chr.12: 4915714-4915714 on Build GRCh38
rs951194413 Chr.12: 4915757-4915757 on Build GRCh38
rs953253014 Chr.12: 4915941-4915941 on Build GRCh38
rs953338389 Chr.12: 4916085-4916085 on Build GRCh38
rs878986774 Chr.12: 4915876-4915876 on Build GRCh38
rs1013376594 Chr.12: 4915908-4915908 on Build GRCh38
rs547570693 Chr.12: 4915853-4915853 on Build GRCh38
rs966516497 Chr.12: 4916066-4916066 on Build GRCh38
rs923945668 Chr.12: 4916008-4916008 on Build GRCh38
rs146125116 Chr.12: 4915790-4915790 on Build GRCh38
rs953380567 Chr.12: 4915677-4915677 on Build GRCh38
rs549320011 Chr.12: 4915923-4915923 on Build GRCh38
rs966176063 Chr.12: 4915807-4915807 on Build GRCh38
rs576248126 Chr.12: 4915715-4915715 on Build GRCh38
rs896251781 Chr.12: 4915666-4915666 on Build GRCh38
rs757025781 Chr.12: 4915815-4915815 on Build GRCh38
rs931980786 Chr.12: 4915841-4915841 on Build GRCh38
rs886049526 Chr.12: 4916051-4916051 on Build GRCh38
rs10849174 Chr.12: 4915928-4915928 on Build GRCh38
rs887424915 Chr.12: 4915819-4915819 on Build GRCh38
rs1056579067 Chr.12: 4916090-4916090 on Build GRCh38
rs1026860543 Chr.12: 4915741-4915741 on Build GRCh38
rs975924632 Chr.12: 4915660-4915660 on Build GRCh38
rs759905728 Chr.12: 4915866-4915866 on Build GRCh38
rs978120428 Chr.12: 4915966-4915966 on Build GRCh38
rs545162947 Chr.12: 4915740-4915740 on Build GRCh38
rs1052162758 Chr.12: 4915657-4915657 on Build GRCh38
rs915198598 Chr.12: 4915686-4915686 on Build GRCh38
rs571352674 Chr.12: 4916070-4916070 on Build GRCh38
rs968038032 Chr.12: 4915963-4915963 on Build GRCh38
rs565320844 Chr.12: 4915749-4915749 on Build GRCh38
rs538124999 Chr.12: 4915949-4915949 on Build GRCh38
rs921987159 Chr.12: 4915827-4915827 on Build GRCh38
rs982590781 Chr.12: 4915779-4915779 on Build GRCh38

View Full Product Details