Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6818740-6819244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD4
Entrez Gene ID
920
Gene Name
CD4 molecule
Gene Aliases
CD4mut
UniGene
Hs.631659
Gene Chromosome Location
Chr.12: 6789472-6820810 on Build GRCh38
Transcripts
NM_000616.4 NM_001195014.2 NM_001195015.2 NM_001195016.2 NM_001195017.2 XM_017020228.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6000531 c.1305C>G p.I435M Chr.12: 6818873-6818873 on Build GRCh38
3765665 c.1341T>C p.C447C Chr.12: 6818909-6818909 on Build GRCh38
942927 c.1279C>T p.R427C Chr.12: 6818847-6818847 on Build GRCh38
3464516 c.1317C>T p.L439L Chr.12: 6818885-6818885 on Build GRCh38
1128482 c.1330A>C p.T444P Chr.12: 6818898-6818898 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782034289 Chr.12: 6819215-6819215 on Build GRCh38
rs782771769 Chr.12: 6819079-6819079 on Build GRCh38
rs782256769 Chr.12: 6818905-6818905 on Build GRCh38
rs199617771 Chr.12: 6818860-6818860 on Build GRCh38
rs782672363 Chr.12: 6818828-6818828 on Build GRCh38
rs782149774 Chr.12: 6818848-6818848 on Build GRCh38
rs782122218 Chr.12: 6818885-6818885 on Build GRCh38
rs781949199 Chr.12: 6818945-6818945 on Build GRCh38
rs782597386 Chr.12: 6818838-6818838 on Build GRCh38
rs575265574 Chr.12: 6818956-6818956 on Build GRCh38
rs781834118 Chr.12: 6818880-6818880 on Build GRCh38
rs782152695 Chr.12: 6818800-6818800 on Build GRCh38
rs782384312 Chr.12: 6818950-6818950 on Build GRCh38
rs782771598 Chr.12: 6818832-6818832 on Build GRCh38
rs782561165 Chr.12: 6818893-6818893 on Build GRCh38
rs200616911 Chr.12: 6818819-6818819 on Build GRCh38
rs782371596 Chr.12: 6818844-6818844 on Build GRCh38
rs782090987 Chr.12: 6818948-6818948 on Build GRCh38
rs56370442 Chr.12: 6818793-6818793 on Build GRCh38
rs781839117 Chr.12: 6818978-6818978 on Build GRCh38
rs782698714 Chr.12: 6818818-6818818 on Build GRCh38
rs942081465 Chr.12: 6819140-6819140 on Build GRCh38
rs561696022 Chr.12: 6819216-6819216 on Build GRCh38
rs782492569 Chr.12: 6818957-6818957 on Build GRCh38
rs28919574 Chr.12: 6818933-6818933 on Build GRCh38
rs2071077 Chr.12: 6818970-6818970 on Build GRCh38
rs1044850684 Chr.12: 6819120-6819120 on Build GRCh38
rs12298133 Chr.12: 6819048-6819048 on Build GRCh38
rs145973129 Chr.12: 6818898-6818898 on Build GRCh38
rs947878884 Chr.12: 6819003-6819003 on Build GRCh38
rs897819888 Chr.12: 6819201-6819201 on Build GRCh38
rs781912242 Chr.12: 6818859-6818859 on Build GRCh38
rs372858201 Chr.12: 6819163-6819163 on Build GRCh38
rs142225058 Chr.12: 6818890-6818890 on Build GRCh38
rs1036414042 Chr.12: 6819179-6819179 on Build GRCh38
rs189212854 Chr.12: 6818964-6818964 on Build GRCh38
rs782581981 Chr.12: 6819184-6819184 on Build GRCh38
rs782569560 Chr.12: 6818894-6818894 on Build GRCh38
rs782252431 Chr.12: 6818950-6818951 on Build GRCh38
rs12821756 Chr.12: 6818909-6818909 on Build GRCh38
rs782632037 Chr.12: 6818959-6818959 on Build GRCh38
rs199710994 Chr.12: 6818808-6818808 on Build GRCh38
rs781997465 Chr.12: 6818847-6818847 on Build GRCh38
rs782528984 Chr.12: 6818822-6818822 on Build GRCh38
rs782308822 Chr.12: 6818835-6818835 on Build GRCh38
rs55764611 Chr.12: 6818969-6818969 on Build GRCh38
rs782716383 Chr.12: 6818878-6818878 on Build GRCh38
rs28919573 Chr.12: 6818843-6818843 on Build GRCh38
rs28919577 Chr.12: 6819017-6819017 on Build GRCh38
rs138218083 Chr.12: 6818854-6818854 on Build GRCh38
rs782783240 Chr.12: 6818888-6818888 on Build GRCh38
rs377662922 Chr.12: 6818820-6818820 on Build GRCh38
rs887632939 Chr.12: 6819127-6819127 on Build GRCh38
rs782395315 Chr.12: 6818791-6818791 on Build GRCh38
rs782261918 Chr.12: 6818952-6818952 on Build GRCh38
rs781847691 Chr.12: 6819183-6819183 on Build GRCh38
rs781808459 Chr.12: 6818897-6818897 on Build GRCh38

View Full Product Details